Biological Buffers
Gly-Gly
Synonym: Diglycine, Glycyl-glycine
Gly-Gly is the simplest of all dipeptides and is used as a starting template for the preparation of more complex peptides1. It is also a substrate for the enzyme glycylglycine dipeptidase2. Valuable agent for experiments investigating peptide transport and absorption.
Useful pH Range 7.5 - 8.9
Working concentration range 120mM3
        Comparison of products by grade or quality level
Name CAS Molecular Formula Mol Wt Reagent Grades Life Science Grades PharmaGrade  Solutions
Gly-Gly 556-50-3 C4H8N2O3 132.12 1.04233 G1002 G3915 50200 50199 G0674  
Interaction Comments
Potential interaction with Zinc and Mercury4
Literature References
  1. The Merck Index, 12th ed., Entry# 4513. 1996
  2. Smith, E. L., Dipeptidases. Methods Enzymol., II, 93-1109 (1955).
  3. Tung K, Mattenheimer H, Burchardt U: Urinary Enzymes in Clinical and Experimental Medicine; Springer-Verlag, 1992
  4. Tewari KC, Lee J, LiNC “Zinc and mercury (II) interaction with cytidine and glycylglycine” Trans. Faraday Soc., 1970,66, 2069-2075