>
Core Bioreagents
>
Biological Buffers
>
MES Sodium Salt
Biological Buffers
MES Sodium Salt
Printer friendly PDF version
(85 Kb)
Sigma-Aldrich
®
MES Sodium Salt Physical Properties
Synonyms
2-(N-Morpholino)ethanesulfonic acid sodium salt
4-Morpholineethanesulfonic acid sodium salt
Molecular Formula
C
6
H
12
NNaO
4
S
Beilstein Registry
3765682
Useful pH range
5.5 - 6.7
Molecular Weight
217.22
MDL Number
MFCD00065473
pKa (at 25)
6.1
CAS Number
71119-23-8
EG/EC Number
275-203-2
Sigma-Aldrich MES Sodium Salt Specifications Comparison
M3885
M5057
M3058
Grade
≥99% (titration)
≥99% (titration)
BPC
Purity
≥99%
≥99%
≥99%
Reagent Testing
M3885
M5057
M3058
Enhanced Quality
Profile Buffers
X
X
X
260NM
<0.05
<0.05
280NM
<0.03
<0.03
290NM
≤0.1
≤0.1
≤0.1
IR
X
X
pH
8.9 - 10.1
8.9 - 10.1
Karl Fischer
≤8%
≤8%
≤8%
Life Science Testing
M3885
M5057
M3058
Synthetic
X
Endotoxin
≤0.1 eu/mg
SPC
≤100 cfu/g
Cell test
X
RNAse
X
DNAse
X
Nickase
X
Protease
X
Lead(Pb)
≤0.0005%
≤0.0005%
≤0.0005%
back to top
back to Biological Buffers
Copyrights © 2010 Sigma-Aldrich Co. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.