>
Core Bioreagents
>
Biological Buffers
>
MES
Biological Buffers
MES
Printer friendly PDF version
(100 Kb)
Sigma-Aldrich
®
MES Physical Properties
Synonyms
2-(N-Morpholino)ethanesulfonic acid
4-Morpholineethanesulfonic acid
Molecular Formula
C
6
H
13
NO
4
S · xH
2
O
CAS Number
4432-31-9
pKa (at 20)
6.16
Molecular Weight
195.24 (anhydrous basis)
Useful pH range
5.5 - 6.7
pKa (at 25)
6.1
pKa (at 37)
5.97
Sigma-Aldrich MES Specifications Comparison
M3671
M8250
M5287
M2933
69890
Grade
low moisture content, ≥99% (titration)
≥99.5% (titration)
≥99.5% (titration), pH 2.5 - 4.0
(20 °C, 0.5 M in H
2
O), SigmaUltra
BPC
, plant cell culture tested, ≥99.5%
BioUltra,
≥99.5% (T)
Purity
≥99%
≥99.5%
≥99.5%
≥99.5%
≥99.5%
Reagent Testing
M3671
M8250
M5287
M2933
69890
Enhanced Quality
Profile Buffers
X
X
X
X
260NM
≤0.025
≤0.025
280NM
≤0.020
≤0.020
290NM
≤0.05
≤0.05
≤0.05
IR
X
X
pH
2.5 - 4.0 (20 °C, 0.5 M in H
2
O)
2.5 - 4.0
(20 °C, 0.5 M in H
2
O)
Karl Fischer
≤1.0%
≤9%
report result
≤9%
Insoluble Matter
filter test
filter test
Residue on Ignition
≤0.05%
≤0.05%
Life Science Testing
M3671
M8250
M5287
M2933
69890
Synthetic
X
Endotoxin
≤0.1 eu/mg
SPC
≤100 cfu/g
Plant cell test
X
RNAse
X
DNAse
X
Nickase
X
Protease
X
Trace Elemental Impurities
M3671
M8250
M5287
M2933
69890
Sodium(Na)
≤0.005%
≤0.0050%
Chloride(Cl)
≤0.005%
≤0.0050%
Sulfate(SO
4
)
≤0.0050%
Aluminum(Al)
≤0.0005%
≤0.0005%
Arsenic(As)
≤0.0001%
≤0.00001%
Barium(Ba)
≤0.0005%
≤0.0005%
Bismuth(Bi)
≤0.0005%
≤0.0005%
Calcium(Ca)
≤0.002%
≤0.0020%
Cadmium(Cd)
≤0.0005%
≤0.0005%
Cobalt(Co)
≤0.0005%
≤0.0005%
Chromium(Cr)
≤0.0005%
≤0.0005%
Copper(Cu)
≤0.0005%
≤0.0005%
Iron(Fe)
≤0.0005%
≤0.0005%
≤0.0005%
Potassium(K)
≤0.005%
≤0.0050%
Lithium(Li)
≤0.0005%
≤0.0005%
Magnesium(Mg)
≤0.0005%
≤0.0005%
Manganese(Mn)
≤0.0005%
≤0.0005%
Molybdenum(Mo)
≤0.0005%
≤0.0005%
Nickel(Ni)
≤0.0005%
≤0.0005%
Lead(Pb)
≤0.0005%
≤0.0005%
≤0.0005%
≤0.0005%
≤0.0005%
Strontium(Sr)
≤0.0005%
≤0.0005%
Zinc(Zn)
≤0.0005%
≤0.0005%
back to top
back to Biological Buffers