|
Biological Buffers
MOPS Sodium Salt
|
|
|
| Sigma-Aldrich® MOPS Sodium Salt Physical Properties |
| Synonyms |
3-(N-Morpholino)propanesulfonic acid sodium salt 4-Morpholinepropanesulfonic acid sodium salt |
 |
| Molecular Formula |
C7H14NNaO4S |
CAS Number |
71119-22-7 |
Useful pH range |
6.5 - 7.9 |
| Molecular Weight |
231.25 |
MDL Number |
MFCD00064350 |
pKa (at 25) |
7.2 |
| Sigma-Aldrich MOPS Sodium Salt Specifications Comparison |
| |
M9381 |
M9024 |
M5789 |
| Grade |
≥99.5% (titration) |
BPC, cell culture tested, ≥99.5% (titration) |
SigmaUltra, ≥99.5% as anhydrous (titration) |
| Purity |
≥99.5% |
≥99.5% |
≥99.5% |
| Reagent Testing |
| |
M9381 |
M9024 |
M5789 |
Enhanced Quality Profile Buffers |
X |
X |
X |
| 260NM |
|
|
≤0.1 |
| 280NM |
|
|
≤0.1 |
| 290NM |
≤0.05 |
≤0.05 |
|
| IR |
X |
X |
|
| pH |
|
|
10.0 - 12.0 (20 °C, 1 M in H2O) |
| Karl Fischer |
≤5% |
≤5% |
≤5% |
| Insoluble Matter |
|
|
≤0.1% |
| Life Science Testing |
| |
M9381 |
M9024 |
M5789 |
| Endotoxin |
|
≤0.1 eu/mg |
|
| SPC |
|
≤100 cfu/g |
|
| Cell test |
|
X |
|
| RNAse |
|
X |
|
| DNAse |
|
X |
|
| Nickase |
|
X |
|
| Protease |
|
X |
|
| Trace Elemental Impurities |
| |
M9381 |
M9024 |
M5789 |
| Ammonia(NH4) |
|
|
≤0.05% |
| Phosphorus(P) |
|
|
≤0.005% |
| Chloride(Cl) |
|
|
≤0.05% |
| Sulfate(SO4) |
|
|
≤0.05% |
| Aluminum(Al) |
|
|
≤0.0005% |
| Calcium(Ca) |
|
|
≤0.001% |
| Copper(Cu) |
|
|
≤0.0005% |
| Iron(Fe) |
|
|
≤0.0005% |
| Potassium(K) |
|
|
≤0.005% |
| Magnesium(Mg) |
|
|
≤0.0005% |
| Lead(Pb) |
≤0.0005% |
≤0.0005% |
≤0.001% |
| Zinc(Zn) |
|
|
≤0.0005% |
| |
back to top back to Biological Buffers |