|
In an effort to provide a product that meets the growing demands of the Life Science market, Sigma-Aldrich has developed the Biotechnology Performance Certified product line. In addition to our regular testing, these products are qualified for Molecular Biology, Tissue Culture, and Electrophoresis (where applicable). The competitively priced Sigma-Aldrich BPC product line will eliminate the need to purchase multiple grades of the same product helping the customer save both dollars and storage space.
Selected Sigma-Aldrich BPC Buffers
|
Product Code
|
Product Name
|
Heavy Metals (PB) |
Fe |
Endotoxin |
SPC1 |
KF |
Dnase, Rnase, Protease free |
Cell2 |
| A3594 |
Aces |
X |
|
X |
X |
|
X |
|
| B4554 |
BES Free Acid |
X |
|
X |
X |
X |
X |
X |
| B4429 |
Bis-Tris Free Base |
X |
|
X |
X |
X |
X |
X |
| B4679 |
Bis-Tris Propane |
X |
|
X |
X |
X |
X |
X |
| B1934 |
Boric Acid |
X |
|
X |
X |
|
X |
X |
| E0276 |
EPPS |
X |
|
X |
X |
X |
X |
X |
| G3915 |
Gly-Gly |
X |
|
X |
X |
|
X |
X |
| H4034 |
HEPES Free Acid |
X |
X |
X |
X |
X |
X |
X |
| H3537 |
HEPES Free Acid 1 M Solution |
X |
|
X |
X |
|
X |
X |
| H3784 |
HEPES Sodium |
X |
X |
X |
X |
X |
X |
X |
| H3662 |
HEPES Sodium 1 M Solution |
X |
|
X |
X |
|
X |
X |
| M2933 |
MES Free Acid |
X |
X |
X |
X |
X |
X |
X |
| H1317 |
MES Free Acid 1 M Solution |
X |
|
X |
X |
|
X |
X |
| M3058 |
MES Sodium Salt |
X |
|
X |
X |
X |
X |
X |
| M1442 |
MOPS 1 M Solution |
X |
|
X |
X |
|
X |
X |
| M3183 |
MOPS Free Acid |
X |
X |
X |
X |
X |
X |
X |
| M9024 |
MOPS Sodium |
X |
|
X |
X |
X |
X |
X |
| P5493 |
Phosphate Buffered Saline |
X |
|
X |
X |
|
X |
X |
| P1851 |
Pipes Free Acid |
X |
|
X |
X |
|
X |
X |
| S3817 |
Sodium Bicarbonate |
X |
|
X |
X |
|
X |
|
| S6191 |
Sodium Chloride |
X |
|
X |
|
|
X |
X |
| S6316 |
Sodium Chloride 5 M Solution |
X |
|
X |
X |
|
X |
X |
| S6566 |
Sodium Phosphate Monobasic Anhy |
X |
X |
X |
X |
X |
X |
X |
| T5316 |
TAPS Free Acid |
X |
X |
X |
X |
X |
X |
|
| T5441 |
TAPS Sodium |
X |
|
X |
X |
X |
X |
X |
| T5566 |
TAPSO Free Acid |
X |
|
X |
X |
X |
X |
X |
| T5691 |
TES Free Acid |
X |
|
X |
X |
X |
X |
X |
| T5816 |
Tricine |
X |
|
X |
X |
X |
X |
X |
| T1074 |
Tricine 1 M Solution |
X |
X |
X |
X |
|
X |
X |
| T6066 |
Trizma Base |
X |
|
X |
X |
X |
X |
X |
| T5941 |
Trizma Hydrochloride |
X |
|
X |
X |
X |
X |
X |
SPC = Standard Plate Count Cell = Cell Culture Test. Product is diluted to a working concentration in cell media and used to grow cells. |
|