|
Biological Buffers
Sodium Chloride
|
|
|
| Sigma-Aldrich® Sodium Chloride Physical Properties |
| Molecular Formula |
NaCl |
Beilstein Registry |
3534976 |
Melting Point |
801 °C(lit.) |
| Molecular Weight |
58.44 |
MDL Number |
MFCD00003477 |
EG/EC Number |
231-598-3 |
| CAS Number |
7647-14-5 |
|
| Sigma-Aldrich Sodium Chloride Specifications Comparison |
| |
S1679 |
S9888 |
S5629 |
S5886 |
S3014 |
S6191 |
S7653 |
71376 |
| Grade |
meets USP testing specifications |
ACS reagent ≥99.0% |
plant cell culture tested, ≥99.5% |
cell, insect cell culture tested |
for molecular biology, ≥98% (titration) |
BPC, ≥99.5% (titration), cell culture tested |
BioXtra, ≥99.5% |
BioUltra, for molecular biology, ≥99.5% (AT) |
| Purity |
≥99.0% |
≥99.0% |
≥99.5% |
≥99.5% |
≥98% |
≥99.5% |
≥99.5% |
≥99.5% |
| Reagent Testing |
| |
S1679 |
S9888 |
S5629 |
S5886 |
S3014 |
S6191 |
S7653 |
71376 |
| 260NM |
|
≤0.01 |
≤0.01 |
| 280NM |
|
≤0.01 |
≤0.01 |
| Identity |
X |
|
| pH |
|
5.0 - 9.0 (25 °C, 5% in solution) |
|
5.0 - 8.0 (20 °C, 1M in H2O) |
5.0 - 8.0 (25 °C, 1M in H2O) |
| LOD |
≤0.5% |
|
| Insoluable Matter |
|
≤0.005% |
|
filter test |
filter test |
| Life Science Testing |
| |
S1679 |
S9888 |
S5629 |
S5886 |
S3014 |
S6191 |
S7653 |
71376 |
| Endotoxin |
|
X |
|
|
| Cell test |
|
|
|
X |
|
X |
|
|
| Insect cell test |
|
|
|
X |
|
X |
|
|
| Plant cell test |
|
|
X |
|
|
X |
|
|
| RNase |
|
X |
X |
|
X |
| DNAse |
|
X |
X |
|
X |
| Protease |
|
X |
X |
|
X |
| Phosphatase |
|
X |
| Trace Elemental Impurities |
| |
S1679 |
S9888 |
S5629 |
S5886 |
S3014 |
S6191 |
S7653 |
71376 |
| Bromide(Br) |
≤0.010% |
≤0.01% |
|
≤0.005% |
≤0.0050% |
| Iodide(I) |
X |
≤0.002% |
|
≤0.001% |
≤0.0010% |
| Phosphate(P) |
≤0.0025% |
≤0.0005% |
|
|
≤0.0005% |
|
≤0.0005% |
≤0.0005% |
| Sulfate(SO4) |
≤0.020% |
≤0.004% |
|
≤0.05% |
≤0.0100% |
| Aluminum(Al) |
|
≤0.0005% |
≤0.0005% |
| Arsenic(As) |
≤0.0001% |
|
≤0.0001% |
≤0.00001% |
| Barium(Ba) |
X |
X |
|
≤0.0005% |
≤0.0005% |
| Bismuth(Bi) |
|
≤0.0005% |
≤0.0005% |
| Calcium(Ca) |
|
≤0.002% |
|
≤0.002% |
≤0.0010% |
| Cadmium(Cd) |
|
≤0.0005% |
≤0.0005% |
| Cobalt(Co) |
|
≤0.0005% |
≤0.0005% |
| Chromium(Cr) |
|
≤0.0005% |
≤0.0005% |
| Copper(Cu) |
|
≤0.0005% |
≤0.0005% |
| Iron(Fe) |
≤0.0002% |
≤0.0002% |
|
≤0.0001% |
≤0.0001% |
| Potassium(K) |
≤0.05% |
≤0.005% |
|
|
≤0.0050% |
|
≤0.005% |
≤0.0050% |
| Lithium(L) |
|
≤0.0005% |
≤0.0005% |
| Magnesium(Mg) |
|
≤0.001% |
|
≤0.0005% |
≤0.0005% |
| Manganese(Mn) |
|
≤0.0005% |
≤0.0005% |
| Molybdenum |
|
≤0.0005% |
≤0.0005% |
| Nickel(Ni) |
|
≤0.0005% |
≤0.0005% |
| Lead(Pb) |
|
≤0.0005% |
|
|
≤0.0005% |
≤0.0005% |
≤0.0005% |
≤0.0005% |
| Strontium(Sr) |
|
≤0.0005% |
≤0.0005% |
| Zinc(Zn) |
|
≤0.0005% |
≤0.0005% |
| Total Nitrogen |
|
≤0.001% |
| Chlorate and Nitrate(NO3) |
|
≤0.003% |
|
| Ferrocyanides |
X |
|
| Magnesium and Alkaline Earth Metals |
≤0.01% |
|
| Nitrites |
X |
|
| Heavy Metals |
≤0.0005% |
|
| |
back to top back to Biological Buffers |