Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Biological Buffers
Tricine
Synonym: N-[Tris(hydroxymethyl)methyl]glycine
Tricine is a zwitterionic amino acid. It is an organic derivative of tris and glycine used in buffer solutions. Tricine has been recommended as the buffer of choice in SDS-PAGE systems, in separating proteins in the range of 1 to 100 kDa1. Tricine is also an effective hydroxyl radical scavenger used to protect molecules during radical generating procedures such as irradiation2.
Useful pH Range 7.4 - 8.8
Working concentration range 50mM3
        Comparison of products by grade or quality level
Name CAS Molecular Formula Mol Wt Reagent Grades Life Science Grades PharmaGrade  Solution
Tricine 5704-04-1 C6H13NO5 179.17  
Interaction Comments
Tricine strongly interacts with Magnesium, Calcium, Copper, Cobalt, Nickel and Zinc5
Literature References
  1. Schäger, H. and von Jagow, G., Anal. Biochem., 166, 368-379 (1987).
  2. Hicks, M. and Gebicki, J.M., FEBS Letters, 199(1), 92 (1986).
  3. Harrison MT et al, 2009 "Nitrogen in cell walls of sclerophyllous leaves accounts for little of the variation in photosynthetic nitrogen-use efficiency" Plant, Cell and Environment (32) 259-270
  4. Sorensen H et al Chromatography and Capillary Electrophoresis in Food Analysis. Royal Soceity of Chemistry, 1999.
  5. Ferreira CM, Pinto IS, Soares, EV, Soares HM. 2015 Royal Society of Chemistry 30989- 31003