|
Trizma® Base (Tris Base)
Trizma®
|
|
|
| Sigma-Aldrich Trizma Base Physical Properties |
| Synonyms |
2-Amino-2-(hydroxymethyl)-1,3-propanediol THAM Tris base Tris(hydroxymethyl)aminomethane Trometamol |
 |
| Molecular Formula |
NH2C(CH2OH)3 |
Beilstein Registry |
741883 |
Useful pH range |
7.0-9.0 |
| Molecular Weight |
121.14 |
MDL Number |
MFCD00004679 |
pKa ( at 20)
|
8.20 |
| CAS Number |
77-86-1 |
EG/EC Number |
201-064-4 |
pKa (at 25) |
8.06 |
| Melting point |
168-172 °C |
Boiling point |
219-220 °C/10 mmHg |
pKa (at 37) |
7.72 |
| Sigma-Aldrich Trizma Base Specifications Comparison |
| |
T1378 |
252859 |
T1503 |
T6687 |
T6791 |
93350 |
T6066 |
93362 |
| Grade |
Crystalline |
ACS-reagent |
Primary Standard and Buffer, Crystalline |
USP |
SigmaUltra |
Analytical, Puriss P.A. |
BPC, Cell Culture, EP, USP, USP/NF |
BioChemika, BioUltra, Molecular Biology |
| Purity |
≥99% |
≥99.8% |
≥99.9% |
≥99.0% |
≥99.9% |
≥99.7% |
≥99.9% |
≥99.8% |
| Reagent Testing |
| |
T1378 |
252859 |
T1503 |
T6687 |
T6791 |
93350 |
T6066 |
93362 |
|
X |
|
X |
|
X |
|
| 230nm |
|
≤0.6 |
|
|
| 260nm |
|
≤ 0.025 |
≤0.05 |
≤0.06 |
≤0.10 |
| 280nm |
|
≤ 0.020 |
≤0.04 |
≤ 0.06 |
≤0.08 |
| 290nm |
≤0.4 |
≤0.2 |
≤0.05 |
|
|
|
≤ 0.05 |
|
| 430nm |
|
≤0.01 |
|
|
| IR |
|
X |
|
| Identity |
|
|
|
X |
|
| Identity A |
|
X |
|
| Identity B |
|
X |
|
| Identity C |
|
X |
|
| Identity EP |
|
X |
|
| Identity USP |
|
X |
|
| Karl Fischer |
≤0.5% |
|
≤0.2% |
|
|
|
≤0.2% |
|
| LOD |
|
|
|
≤1.0% |
≤0.5% |
|
≤0.5% |
≤0.5% |
| Insoluble Matter |
|
≤0.005% |
|
|
≤0.005% |
|
|
Filter test |
| Residue on Ignition |
|
|
|
≤0.1% |
≤0.01% |
≤0.01% |
≤0.1% |
≤0.01% |
| Titration |
|
≤2% H2O |
|
| Life Science Testing |
| |
T1378 |
252859 |
T1503 |
T6687 |
T6791 |
93350 |
T6066 |
93362 |
| Endotoxin |
|
≤1 eu/mg |
|
| SPC |
|
≤100 cfu/g |
|
| Cell test |
|
x |
|
| RNAse |
|
x |
x |
| DNAse |
|
x |
x |
| Nickase |
|
x |
|
| Protease |
|
x |
x |
| Phosphatase |
|
x |
| Suitability Electrophoresis |
|
x |
|
| Trace Elemental Analysis |
| |
T1378 |
252859 |
T1503 |
T6687 |
T6791 |
93350 |
T6066 |
93362 |
| Sodium(Na) |
|
≤0.0050% |
≤0.0050% |
|
≤0.0050% |
| Chloride(Cl) |
|
≤0.0050% |
≤0.0005% |
|
≤0.0020% |
| Sulfate(SO4) |
|
≤0.0050% |
≤0.0005% |
|
≤0.0005% |
| Aluminum(Al) |
|
≤0.0005% |
|
|
≤0.0005% |
| Arsenic(As) |
|
≤0.00001% |
|
|
≤0.00001% |
| Barium(Ba) |
|
≤0.0005% |
|
|
≤0.0005% |
| Bismuth(Bi) |
|
≤0.0005% |
|
|
≤0.0005% |
| Calcium(Ca) |
|
≤0.0010% |
≤0.0010% |
|
≤0.0010% |
| Cadmium(Cd) |
|
≤0.0005% |
≤0.0005% |
|
≤0.0005% |
| Cobalt(Co) |
|
≤0.0005% |
≤0.0005% |
|
≤0.0005% |
| Chromium(Cr) |
|
≤0.0005% |
≤0.0005% |
|
≤0.0005% |
| Copper(Cu) |
|
≤0.0005% |
≤0.0005% |
|
≤0.0005% |
| Iron(Fe) |
|
≤0.0005% |
|
|
≤0.0005% |
≤0.0005% |
≤0.0001% |
≤0.0005% |
| Potassium(K) |
|
≤0.0050% |
≤0.0050% |
|
≤0.0050% |
| Lithium(Li) |
|
≤0.0005% |
|
|
≤0.0005% |
| Magnesium(Mg) |
|
≤0.0005% |
≤0.0005% |
|
≤0.0005% |
| Manganese(Mn) |
|
≤0.0005% |
≤0.0005% |
|
≤0.0005% |
| Molybdenum(Mo) |
|
≤0.0005% |
|
|
≤0.0005% |
| Nickel(Ni) |
|
≤0.0005% |
≤0.0005% |
|
≤0.0005% |
| Lead(Pb) |
≤0.0005% |
≤0.0005% |
≤0.0002% |
|
≤0.0005% |
≤0.0005% |
≤0.0002% |
≤0.0005% |
| Strontium(Sr) |
|
≤0.0005% |
|
|
≤0.0005% |
| Zinc(Zn) |
|
≤0.00005% |
≤ 0.0005% |
|
≤ 0.0005% |
| Related substances |
|
x |
|
| Heavy Metals |
|
|
|
≤0.001% |
|
| Chlorides |
|
≤0.0100% |
|
| |
| back to top |
Back to Biological Buffers
|