|
Core Bioreagents
Technical Library
|
Buffer pKa/pH Tables and Formulas: Useful pH Ranges of Selected Biological Buffers (25 °C, 0.1M) Trizma Buffer Table - pH vs. Temperature Phosphate Buffer Table - 0.2M solution Commonly Used Electrophoresis Buffers Helpful Formulas
DNA / Protein Electrophoresis and Troubleshooting Tables: Effective Range of Separation of DNA in Polyacrylamide Gels Effective Range of Separation of DNA in Agarose Gels DNA Size Migration of Sample Loading Dyes Troubleshooting Guide for SDS-PAGE Protein Electrophoresis
Genetic Markers Tables: Significant Genetic Markers in Commonly Used E. coli Strains Other Genetic Markers that Affect Growth Requirements in Commonly Used E. coli Strains
RNase Contamination Prevention: Quick Reference Guide for Preventing RNase Contamination
Nucleotide / Amino Acids Properties: Nucleotide Physical Properties Amino Acids Table
Life Science Conversions: Mass to Moles Conversions Molecular Masses of Nucleic Acids Protein Conversion Data Concentration of Protein to Absorbance of Protein Mass of Protein to Mole of Protein
Buffer pKa/pH Tables and Formulas:
Useful pH Ranges of Selected Biological Buffers (25 °C, 0.1M)
Trizma Buffer Table - pH vs. Temperature
| pH at Temperature |
|
g/L for 0.05 M Solution |
| 5°C |
25°C |
37°C |
|
Trizma HCL |
Trizma Base |
| 7.76 |
7.20 |
6.91 |
|
7.02 |
0.67 |
| 7.89 |
7.30 |
7.02 |
|
6.85 |
0.80 |
| 7.97 |
7.40 |
7.12 |
|
6.61 |
0.97 |
| 8.07 |
7.50 |
7.22 |
|
6.35 |
1.18 |
| 8.26 |
7.70 |
7.40 |
|
5.72 |
1.66 |
| 8.37 |
7.80 |
7.52 |
|
5.32 |
1.97 |
| 8.48 |
7.90 |
7.62 |
|
4.88 |
2.30 |
| 8.58 |
8.00 |
7.71 |
|
4.44 |
2.65 |
| 8.68 |
8.10 |
7.80 |
|
4.02 |
2.97 |
| 8.78 |
8.20 |
7.91 |
|
3.54 |
3.34 |
| 8.88 |
8.30 |
8.01 |
|
3.07 |
3.70 |
| 8.98 |
8.40 |
8.10 |
|
2.64 |
4.03 |
| 9.09 |
8.50 |
8.22 |
|
2.21 |
4.36 |
| 9.18 |
8.60 |
8.31 |
|
1.83 |
4.65 |
| 9.28 |
8.70 |
8.42 |
|
1.50 |
4.90 |
| 9.36 |
8.80 |
8.51 |
|
1.23 |
5.13 |
| 9.47 |
8.90 |
8.62 |
|
0.96 |
5.32 |
| 9.56 |
9.00 |
8.70 |
|
0.76 |
5.47 |
| |
back to top |
Phosphate Buffer Table - 0.2M solution
| Sodium Phosphate Monobasic Anhydrous g/L |
Sodium Phosphate Dibasic Heptahydrate g/L |
23°C pH |
|
Sodium Phosphate Monobasic Anhydrous g/L |
Sodium Phosphate Dibasic Heptahydrate g/L |
23°C pH |
| 22.4 |
3.49 |
5.7 |
|
10.80 |
29.51 |
6.9 |
| 22.08 |
4.29 |
5.8 |
|
9.36 |
32.73 |
7.0 |
| 21.60 |
5.37 |
5.9 |
|
7.92 |
35.95 |
7.1 |
| 21.05 |
6.60 |
6.0 |
|
6.72 |
38.63 |
7.2 |
| 20.40 |
8.05 |
6.1 |
|
5.52 |
41.31 |
7.3 |
| 19.56 |
9.93 |
6.2 |
|
4.56 |
43.46 |
7.4 |
| 18.60 |
12.07 |
6.3 |
|
3.84 |
45.07 |
7.5 |
| 17.64 |
14.22 |
6.4 |
|
3.12 |
46.68 |
7.6 |
| 16.44 |
16.90 |
6.5 |
|
2.52 |
48.55 |
7.7 |
| 15.00 |
20.12 |
6.6 |
|
2.04 |
49.09 |
7.8 |
| 13.56 |
23.34 |
6.7 |
|
1.68 |
49.89 |
7.9 |
| 12.24 |
26.29 |
6.8 |
|
1.27 |
50.81 |
8.0 |
| |
back to top |
Commonly Used Electrophoresis Buffers |
| Tris-Acetate (TAE) |
1X |
0.4 M Tris-Acetate/0.001 M EDTA, pH 8.3 |
| Tris-Phosphate (TPE) |
1X |
0.08 M Tris-Phosphate/0.002 M EDTA, pH 8.0 |
| Tris-Borate (TBE) |
1X |
0.089 M Tris-Borate/0.002 M EDTA, pH 8.3 |
Helpful Formulas
Percentage by weight (w/v) (% buffer desired / 100) x final buffer volume (ml) = g of starting material needed. |
Molar Solutions desired molarity x formula weight x solution final volume (L) = grams needed |
DNA / Protein Electrophoresis and Troubleshooting Tables
| Effective Range of Separation of DNA in Polyacrylamide Gels |
| % Acrylamide (w/v)¹ |
Efficient Range of Separation (bp) |
| 3.5 |
1000-2000 |
| 5.0 |
80-500 |
| 8.0 |
60-400 |
| 12.0 |
40-200 |
| 15.0 |
25-150 |
| 20.0 |
6-100 |
| 1) N,N'-methylenebisacrylamide is included at 1/30th the concentration of acrylamide. |
Effective Range of Separation of DNA in Agarose Gels |
| % Agarose (w/v) |
Efficient Range of Separation of Linear DNA Molecules (kb) |
| 0.3 |
5-60 |
| 0.6 |
1-20 |
| 0.7 |
0.8-10 |
| 0.9 |
0.5-7 |
| 1.2 |
0.4-6 |
| 1.5 |
0.2-3 |
| 2.0 |
0.1-2 |
DNA Size Migration of Sample Loading Dyes |
| Agarose Concentration (%w/v) |
Xylene Cyanole |
Bromophenol Blue |
| 0.1-1.5 |
4-5 kb |
400-500 bp |
| 2.0-3.0 (sieving agarose) |
750 bp |
100 bp |
| 4.0-5.0 (sieving agarose) |
125 bp |
25 bp |
Troubleshooting Guide for SDS-PAGE Protein Electrophoresis |
| Problem |
Possible causes |
Solution |
| Faint or missing protein bands |
Load quantity is below the detection level of the stain |
Check the A280 and increase sample concentration |
| |
|
Use a more sensitive stain (e.g. silver stain) |
| |
Proteins were not fixed in the gel |
Use a stain which also fixes the proteins |
| |
|
Use gel fixing solution |
| |
Small peptides (<4 kDa) did not fix in the gel |
Fix the gel with 5% glutaraldehyde. Rinse the gel well with water before staining |
| |
Proteins are degraded |
Check the A280 and avoid protease contamination Avoid freeze-thaw of samples |
| |
Protein ran off the gel |
Use a higher concentration PAGE gel. See precast gels for recommended gel concentration or use a 4-20% gel if the size is unknown |
| Film on gel after staining |
Precipitated Coomassie Blue R |
Rinse the gel for 15 seconds in methanol and immediately return to water or destain |
| Poor band resolution |
Concentration of protein is high |
Load 10 μg per protein or 100 μg per protein extract |
| |
Age of the gel, due to base catalyzed hydrolysis of the amide |
Order fresh precast gels or cast a fresh gel |
| |
Improper gel concentration |
See precast gels for recommended gel concentration or use a 4-20% gel if the size is unknown |
| Band smearing |
High salt concentrations |
Dialyze sample, precipitate the protein with TCA or use desalting columns |
| |
Concentration of protein is high |
Load 10 μg per protein or 100 μg per protein extract |
| |
Protein aggregation |
Add 4-8 M urea to the sample Add fresh DTT (30 mM) or 2-mercaptoethanol (5%) |
| |
Voltage is high |
Electrophorese at 10-15 V/cm |
| Protein precipitation in the well |
Hydrophobic proteins |
Add 4-8 M urea to the sample |
| White precipitate in sample |
SDS precipitation |
Could be due to the presence of guanidine or potassium salts in the sample |
Genetic Markers Tables
| Significant Genetic Markers in Commonly Used E. coli Strains |
| Marker |
Description |
Significance |
| dam |
Endogenous adenine methylation at GATC sequences abolished |
High recombination frequency, constitutively express DNA repair functions. Makes DNA susceptible to cleavage by certain restriction enzymes |
| deoR |
Regulatory gene that allows for constitutive synthesis of genes involved in deoxyribose synthesis |
Allows the uptake of large plasmids |
| dnaj |
One of several chaperonins inactive |
Stabilizes certain mutant proteins expressed in E.coli |
| endA1 |
Endonuclease mutation |
Absence of endonuclease improves the quality and yield of plasmid DNA |
| F' |
F' episome, male E. coli host |
Necessary for M13 infection |
| gyrA |
Mutation in DNA gyrase |
Confers resistance to nalidixic acid |
| laclq |
Overproduces the lac repressor |
Overproduces the lac repressor protein, which regulates transcription from the lac promoter |
| lacZΔM15 |
Partial deletion of lacZ that allows α-complementation |
Required for use with pUC or M13 vectors. Results in blue and white colonies or plaques for clone selection when plated on X-gal |
| ΔIon |
Deletion of the Ion protease |
Reduces degradation of β-galactosidase fusion proteins to enhance antibody screening of φ libraries |
| mcrB |
Mutation eliminating restriction of DNA methylated at the sequence 5'-GmC-3' |
Absence of the mcrB gene product allows more efficient cloning of DNA containing 5-methylcytosine or 5-hydroxymethylcytosine |
| mrr |
Mutation eliminating restriction of DNA methylated at the sequence 5'-CmAG-3' |
Absence of the mrr gene product allows more efficient cloning of DNA containing methyladenine residues |
| recA |
Homologous recombination abolished |
Useful when working with sequences containing direct repeats >50 bp |
| recB, recC |
Exonuclease and recombination activity of Exonuclease V abolished |
Stability of inverted repeat sequences enhanced |
| recD |
Exonuclease activity of ExoV abolished |
Inverted repeat sequences in λ can be propagated |
| supE/supF |
Amber suppressors |
Needed to grow amber mutants |
| Tn5 |
Transposon |
Encodes resistance to the antibiotic kanamycin |
| Tn10 |
Transposon |
Encodes resistance to the antibiotic tetracycline |
Other Genetic Markers that Affect Growth Requirements in Commonly used E. coli Strains |
| Marker |
Effect on Growth |
| IeuB |
Requires leucine |
| metB |
Requires methionine |
| proA/B |
Requires proline |
| thi-1 |
Requires thiamine |
| galK/U |
Cannot metabolize galactose |
| mtlA |
Cannot metabolize mannitol |
| xyl5 |
Cannot metabolize xylose |
| nupG |
Alters nucleoside uptake |
RNase Contamination Prevention
| Quick Reference Guide for Preventing RNase Contamination |
| Precautions |
Methods |
Reason |
Hints |
| General lab space |
See below |
To create an RNase-free area in which to work with RNA |
All equipment in this area should be RNase-free. Everything should be handled with gloves and wiped down with 70% ethanol and DEPC treated water before and after use. |
| Equipment |
Wiped down with 70% ethanol and DEPC treated water |
To prevent contamination of samples from local and airborne RNases |
All equipment in this area should be RNase free. Everything should be handled with gloves and wiped down with 70% ethanol and DEPC treated water before and after use. |
| Gloves |
|
RNase present in oil on hands |
Change gloves often as doorknobs, micropipettors, and refrigerator door handles may be contaminated. |
| Glass |
Bake in a dry heat oven 3-4 hours at 180-200 °C |
Inactivates RNase |
Effective method of purging glassware of RNase activity. Some labs set aside equipment that will be used exclusively for RNA work: gel boxes, pipettors, and glassware. |
| Metalware |
Bake in a dry heat oven 3-4 hours at 180-200 °C |
|
Effective method of purging heat resistant equipment of RNase activity. |
| Plasticware |
Autoclave in small batches for RNA use only |
Continuous handling of plasticware can lead to RNase contamination |
Use individually wrapped sterile products. Always handle RNase free tubes (conical and microcentrifuge) with gloves. |
Polycarbonate and polystyrene equipment |
3% hydrogen peroxide |
|
Soak in a 3% hydrogen peroxide solution for 10 minutes and then rinse thoroughly with RNase free water (see below). |
| Solutions |
DEPC |
Chemical RNase inhibitor (not compatible with polycarbonate or polystyrene) |
Use at 0.1% in water. Incubate for several hours and autoclave after treatment in order to destroy remaining DEPC. (Note: Do not add DEPC to any buffers containing Trizma or mercaptans. DEPC is reactive with these products. Instead use DEPC treated water to make up Trizma containing buffers.) |
1. Farrell, R.E. (1993) RNA Methodologies: A Laboratory Guide for Isolation and Characterization. Academic Press, San Diego, CA 33-39 2. Blumberg, D.D., Creating a ribonuclease-free environment. Methods in Enzymol., 152, 20-24 (1987). 3. Sambrook, J., et al., Molecular Cloning: A Laboratory Manual. Cold Spring Harbor Press, Cold Spring Harbor, NY. |
Nucleotide / Amino Acids Properites
Nucleotide Physical Properties |
| Nucleotide |
Molecular Weight (Free Acid) |
λmax (pH 7) |
εm at λmax (pH 7) |
| ATP |
507.2 |
259 |
15,400 |
| CTP |
483.2 |
271 |
9,000 |
| GTP |
523.2 |
253 |
13,700 |
| UTP |
484.2 |
262 |
10,000 |
| dATP |
491.2 |
260 |
15,100 |
| dCTP |
467.2 |
270 |
9,100 |
| dGTP |
507.2 |
253 |
13,800* |
| TTP |
482.2 |
267 |
9,600 |
*εm value for deoxyguanosine monophosphate (dGMP) Reference: Specifications and Criteria for Biochemical Compounds, 3 Ed. Sigma Chemical Company (St. Louis, MO: 1984) |
| Amino Acids Table |
| Amino Acid |
Three-Letter Code |
Single-Letter Code |
Formula |
MW |
| Alanine |
Ala |
A |
C3H7NO2 |
89.1 |
| Arginine |
Arg |
R |
C6H14N4O2 |
174.2 |
| Asparagine |
Asn |
N |
C4H8N2O3 |
132.1 |
| Aspartic Acid |
Asp |
D |
C4H7NO4 |
133.1 |
| Cysteine |
Cys |
C |
C3H7NO2S |
121.2 |
| Glutamic Acid |
Glu |
E |
C5H9NO4 |
147.1 |
| Glutamine |
Gln |
Q |
C5H10N2O3 |
146.1 |
| Glycine |
Gly |
G |
C2H5NO2 |
75.1 |
| Histidine |
His |
H |
C6H9N3O2 |
155.2 |
| Isoleucine |
Ile |
I |
C6H13NO2 |
131.2 |
| Leucine |
Leu |
L |
C6H13NO2 |
131.2 |
| Lysine |
Lys |
K |
C6H14N2O2 |
146.2 |
| Methionine |
Met |
M |
C5H11NO2S |
149.2 |
| Phenylalanine |
Phe |
F |
C9H11NO2 |
165.2 |
| Proline |
Pro |
P |
C5H9NO2 |
115.2 |
| Serine |
Ser |
S |
C3H7NO3 |
105.1 |
| Thereonine |
Thr |
T |
C4H9NO3 |
119.1 |
| Tryptophan |
Trp |
W |
C11H12N2O2 |
204.2 |
| Tyrosine |
Tyr |
Y |
C9H11NO3 |
181.2 |
| Valine |
Val |
V |
C5H11NO2 |
117.1 |
Life Science Conversions
| Mass to Moles Conversions |
| 1 kb DNA Fragment |
|
| 1 μg/ml of DNA |
3.08 μM Phosphate |
| 1 μg/ml of a 1 kb DNA fragment |
3.08 μM 5'-ends |
| 1 μg of a 1 kb DNA fragment |
1.5 pmole = 9.1 x 1011 molecules, 3.0 pmole ends |
| 1 pmole of a 1 kb DNA fragment |
0.65 μg |
| pUC18/19 |
| 1 μg of pUC18/19 DNA (2686 bp) |
0.57 pmole = 3.4 x 1011 molecules |
| 1 pmole of pUC18/19 DNA |
1.77 μ |
| pBR322 DNA |
| 1 μg pBR322 DNA (4361 bp) |
0.35 pmole = 2.1 x 1011 molecules |
| 1 μg of linear pBR322 DNA |
0.70 pmole 5'-ends |
| 1 pmole of pBR322 DNA |
2.83 μg |
| 1 pmole of 5'-ends of linear pBR322 |
1.4 μg |
| M13mp18/19 DNA |
| 1 μg of M13mp18/19 DNA (7249 bp) |
0.21 pmole = 1.3 x 1011 molecules |
| 1 pmole of M13mp18/19 DNA |
4.78 μ |
| λ DNA |
| 1 μg of λ DNA (48,502 bp) |
0.033 pmole = 1.8 x 1010 molecules |
| 1 pmole of λ DNA |
32.01 μ |
| Molecular Masses of Nucleic Acids |
| Average molecular mass of a deoxynucleotide base |
324.5 g/mole |
| Average molecular mass of a deoxynucleotide base pair |
649.0 g/mole1 |
| Average molecular mass of a ribonucleotide base |
340.5 g/mole |
| 1 kb of dsDNA (sodium salt) |
6.5 x 105 g/mole |
| 1 kb of ssDNA (sodium salt) |
3.3 x 105 g/mole |
| 1 kb of ssRNA (sodium salt) |
3.4 x 105 g/mole |
| λ DNA |
3.1 x 107 g/mole2 |
| pBR322 DNA |
2.8 x 106 g/mole2 |
| E.coli DNA |
3.1 x 109 g/mole2 |
| φX174 DNA |
3.6 x 106 g/mole2 |
| 1 x 106 g/mole of dsDNA (sodium salt) |
1.54 kb |
References:
- Ausbel. F.Ml, et al., (ed.), Short Protocols in Molecular Biology, Wiley and Sons, Inc. Ny (1999) pp. A2-1
- Ausbel. F.M., et al., (ed.), Current Protocols in Molecular Biology, Wiley and Sons, Inc. Ny (1999) pp. A.1B1.
|
| Protein Conversion Data Concentration of Protein to Absorbance of Protein |
| Protein |
A280 for 1 mg/ml |
| IgG |
1.35 |
| IgM |
1.20 |
| IgA |
1.30 |
| Protein A |
0.17 |
| Avidin |
1.50 |
| Streptavidin |
3.40 |
| Bovine Serum Albumin |
0.70 |
| Keyhole Limpet Hemocyanin |
1.57 |
| Mass of Protein to Mole of Protein |
| M (g/mole) |
1 μg |
1 nmol |
| 10,000 |
100 pmole; 6 x 1013 molecules |
10 μg |
| 25,000 |
40 pmole; 2.4 x 1013 molecules |
25 μg |
| 50,000 |
20 pmole; 1.2 x 1013 molecules |
50 μg |
| 75,000 |
13.3 pmole; 8 x 1012 molecules |
75 μg |
| 100,000 |
10 pmole; 6 x 1012 molecules |
100 μg |
| 125,000 |
8 pmole; 4.8 x 1012 molecules |
125 μg |
| 150,000 |
6.7 pmole; 4 x 1012 molecules |
150 μg |