Sigma-Aldrich
Life Science > Custom Oligos > Custom DNA Oligos > Learning Center > Modification Yield Chart
Custom DNA Oligos
Modification Yield Chart
Standard | Modifed | S-oligos
Standard Oligos (O.D./µg)*
Scale (µmole) Desalt Cartridge HPLC PAGE
0.03 (3 O.D. Select) 3/90 NA NA NA
0.05 5/150 1.0/30 1.0/30 0.5/15
0.2 12/360 3.0/90 2.5/75 1.0/30
1.0 40/1,200 12/360 13/390 5/150
10 400/12,000 NA 130/3,900 NA
15 600/18,000 NA 190/5,700 NA
* Guarantee is for 20 mers or longer. Shorter oligos may have fewer ODs.
Modified Oligos (O.D./µg)*
Scale (µmole) Desalt Cartridge HPLC PAGE
0.05 2/60 0.4/12 0.4/12 0.2/6
0.2 5/150 1/30 1/30 0.4/12
1.0 16/480 5/150 5/150 2/60
* Guarantee is for 20 mers or longer. Shorter oligos may have fewer ODs. Note: Post-synthesis modifications may yield 50% less than the above stated values.
back to top
Phosphorothioates (S-Oligos) (O.D./µg )*
Scale (µmole) Desalt Cartridge HPLC PAGE
0.2 8.4/252 2.1/63 1.75/52.5 .7/21
1.0 28/840 8.4/252 9.1/273 3.5/105
10 280/8,400 NA 91/2,730 NA
15 420/12,600 NA 133/3990 NA
* Guarantee is for 20 mers or longer. Shorter oligos may have fewer ODs.