Sigma-Aldrich
Life Science > Custom Oligos > Custom DNA Oligos > Learning Center > Oligo Quantification
Custom DNA Oligos
Oligo Quantification
DNA absorbs ultraviolet light due to its highly conjµgated nature. DNA may thus be easily quantitated in a UV spectrometer. Typically, 1 OD 260 (i.e. a solution having an absorbance of one unit at 260 nm with a path length of 1 cm) is said to correspond to a concentration of 30-37 µg per ml. This is for single stranded DNA with an equal mixture of each of the four bases. RNA or double stranded DNA have values of 40 and 50 µg per ml, respectively. For work with DNA longer than probes and primers, these assumptions are valid because the constituent bases are usually fairly evenly represented. With oligos, however, the base composition may be highly skewed. For instance, 1 OD of a sequence of all cytidines would correspond to 39.4 µg/ml and 1 OD of a sequence of all adenosites would correspond to 22.8 µg/ml. Since the composition and sequence of the oligo are usually known, this information may be used to calculate individualized values for more accurate quantitation.
Equations
Values are calculated by either of two similar methods. The following equations is the simplest:
 1.   µg                           1          
-- = --------------------------------------------------
ml (15400A+7400C+11500G+8700T)
------------------------------
1000*(312A+288C+328G+303T-61)
A, C, G, and T are simply the number of each base in the oligo. This equation simply averages the extinction coefficients (in L/mol-Table 1) of each base and divides by the molecular weight (g/mol).
Table1.
Nucleotide Extinction Coefficients at 260 nm
A C G T
15,400 7,400 11,500 8,700
The previous equation ignores the interactions of adjacent bases and a more accurate equation includes nearest neighbor interactions:
2.   µg                           1          
-- = --------------------------------------------------
ml 2*Sum(Eab)-(E2+E3+...+E(n-2)+E(n-1))
--------------------------------------
1000*(312A+288C+328G+303T-61)
Where sum(Eab) is the sum of the pairwise extinction coefficients (Table 2) and the E values are the normal single extinction coefficients for each individual base. (Please note that the first and last bases' extinction coefficients, E1 and E2, are not subtracted.) The pairwise values can be found in Table 2 Both of these equations give good results. The difference between them is rarely more than a few µg per mil.
Table 2. Pairwise extinction coefficients
3' Base
A C G T
5' Base A 13,700 10,600 12,500 11,400
C 10,600 7,300 9,000 7,600
G 12,600 8,800 10,800 10,000
T 11,700 8,100 9,500 8,400
Sigma-Genosys uses equation 2 to determine the extinction coefficient of your oligo.
Examples
For the sequence AGAG, equation (1) gives:
Equation 1 Gives:
     µg                           1          
-- = --------------------------------------------------
ml (15400*2+11500*2)
---------------------
1000*(312*2+328*2-61)
= 22.7
Equation 2 Gives:
µg 1
-- = --------------------------------------------------
ml 2(12500+12600+12500)-(11500+15400)
---------------------------------------
1000*(312*2+328*2-61)
= 25.4
Conclusion
In our example, the results are similar to each other and quite different from the all-purpose value of 30 µg/ml. The degree of accuracy offered by the above equations may not be needed in normal circumstances. Where highly precise quantitation is required, they will yield excellent results.
Sigma-Genosys Guaranteed Yields
Please see the Sigma-Genosys Yields tech sheet for a listing of the yield guarantee for various oligos ordered from Sigma-Genosys.