Sigma-Aldrich
Life Science > Custom Oligos > Custom DNA Oligos > Learning Center > Handling Guidelines for DNA and RNA Oligonucleotides
Custom DNA Oligos
Handling Guidelines for DNA and RNA Oligonucleotides
Oligonucleotides (oligos) require certain basic storage and handling techniques in order to ensure trouble-free experiments. Proper storage of your oligo will maximize its shelf life, allowing you to get the most use from the oligo. These guidelines will enable you to work easily with your oligo in your application.
  • Oligo Resuspension and Storage
  • Preparation of a Concentrated Stock Solution
  • Fluorescently Labeled Oligos
  • Oligonucleotide Quantification
  • Sample Research Application
  • Handling Guidelines for DNA and RNA Oligonucleotides (233 Kb PDF)
Oligo Resuspension and Storage
All DNA oligonucleotides that are provided dry are ready for use upon resuspension, with the exception of Thiol modified oligos. We recommend resuspending oligos in a weak buffer such as TE buffer (10 mM Tris, pH 7.5 – 8.0, 1 mM EDTA, diluted from Product No. T9285). In applications in which TE is not suitable, sterile nuclease-free water (Product No. W4502) may be used. However, high grade water may be slightly acidic and is not recommended for long-term storage. In RNA-related applications, DEPC treated water may be used, but residual DEPC may inhibit subsequent enzymatic reactions.
siRNA duplexes from Sigma® should be resuspended in sterile, nuclease-free water (Product No. W4502). It is not necessary to add buffers when resuspending siRNAs, and after resuspension the siRNAs are ready-to-use.
Single-stranded RNA oligos should be resuspended as decribed above for DNA oligos.
back to top
Preparation of a Concentrated Stock Solution
For all oligos a 100 µMolar stock solution may be obtained by using the following guideline. Take the number of nanomoles (nm) provided (information found on the tube label and/or quality assurance document) and multiply the number of nanomoles by ten. The result provides the number of microliters of liquid (TE, Tris, water or other buffer as required) to add to the tube for reaching a final concentration of 100 µMolar. Note that this is equivalent to a stock solution of 100 pmol/µL. The stock solution may then be further diluted as necessary based upon the application requirements. Store the stock solution at –20 °C in a non-frost-free freezer, and avoid multiple freeze-thaw cycles. Single-stranded RNA oligos should be stored at –80 °C.
back to top
Fluorescently Labeled Oligos
If using fluorescently modified oligos, it is important to note that the intensity of a fluorescent signal can often be directly affected by the pH of the solution used for resuspending. A buffered solution such as TE is recommended for the resuspension of fluorescently labeled oligos. Fluorescently labeled oligos should have minimal exposure to light to prevent dye photobleaching. Wrapping the tubes in foil and/or storing in amber colored tubes is suggested.
back to top
Oligonucleotide Quantification
The microgram quantity of an oligonucleotide is directly related to its ultraviolet absorbance or optical density (OD) at 260 nm. We take great care to ensure that the OD reading determined for each oligo is accurate. You may choose to re-quantify your oligo for verification prior to use. There is a wide range of devices that might be used to measure absorbance at 260 nm. If using a traditional cuvette-based spectrophotometer, the following procedure applies.
Resuspend the oligonucleotide in 400 µl of water. Remove a 12 µl aliquot (to ensure the volume is within the accurate and reproducible range of micropipettes) from the resuspension and dilute with 988 µl of sterile water. Ensure that the OD reading is within the linear range of accuracy of the spectophotometer (typically between 0.1 and 1.5 OD units). If the value is outside the linear range, remeasure using a more appropriate dilution for that sample.
We recommend trying 12 µl from the original 400 µl suspension; therefore, the OD of the 12 µl must be multiplied by a dilution factor of 33 (which is 400/12) and then by the original volume in mL (0.400) to determine the OD of the entire oligo sample.
Sigma calculates µg/OD (A260) based on the extinction coefficient of each base in the oligonucleotide sequence using a nearest-neighbor method. This value is printed on the oligo tube label and QC document that accompanies each shipment. The total number of micrograms of oligo in the 400 µl suspension can be calculated as follows:
Total µg of oligo in original 400 µl = OD reading for 12 µl aliquot x dilution factor 33 x X µg/OD (see tube label)
The quantity of oligonucleotide supplied is expressed in nanomoles on the oligo tube label, along with the µg quantity of the oligo and its molecular weight (MW). Sigma calculates the number of nanomoles using the amount of oligo (in µg) and the molecular weight as follows:
Total nmol = µg of oligo x 1 µmol/MW x 1000 nmol/1 µmol
(Note: 1 mole = 103 mmoles = 106 µmoles = 109 nmoles = 1012 pmoles)
back to top
Sample Research Application
"My tube contains 35.22 nmoles of oligo, but I need a final oligonucleotide concentration of 0.5 uM in my reaction. My reaction volume is 100 ul. How much oligo do I add to my reaction?"
First, make a concentrated oligonucleotide stock solution (10X or 100X, depending on the concentration you need). Then add an appropriate volume to the reaction, thereby achieving the desired working concentration. Serial dilutions of the stock(s) may be required to ensure pipetting accuracy. See outlined example below:
1. Decide on a stock concentration and standardize your units.
Desired final concentration = 0.5 uM
100X stock concentration = 100 X 0.5 uM = 50 micromole/liter
Total nanomoles of oligo = 35.22 nmole (see tube label for your actual oligo)
Total micromoles of oligo = 35.22/1000 = 0.03522 micromole
Note: The quantity of oligo must be expressed in micromoles to calculate the volume for a micromolar solution.
2. Make a 100X stock solution (100 X 0.5 uM = 50 uM).
What volume (V) is required to resuspend 0.03522 micromole of oligo to give a stock concentration of 50 uM? Use the following steps as a guide:
50 micromole
1000 ml
= 0.03522 micromole
V ml
    Solve for V
V = 0.03522 micromole X 1000
50
V = 0.704 ml = 704 ul
3. Use serial dilutions to make a 10X stock (10X 0.5 uM = 5uM).
For pipetting accuracy, avoid using small volumes (less than 2 ul) of stock solutions in your reactions. It may be necessary, therefore, to serially dilute your stock oligonucleotide solution. Make a 10X stock by diluting your 100X stock as follows:
1:10 dilution of 100X stock
10 ul of 50 uM (100X) stock + 90 ul sterile water (or TE) = 100 ul of 5 uM (10X) stock
4. Use a 10X stock to give the final concentration in the reaction.
My reaction volume is 100 ul. How much of the 10X (5 uM) stock solution of oligo is required to yield a 0.5 uM concentration of oligo in the total reaction?
Vol. of (10X) stock X stock conc. = Final reaction vol. X final oligo conc.
(S ul) X (5 uM) = (100 ul) X (0.5 uM) Solve for S
S = 100 X 0.5 ul
5
S = 10 ul