Custom DNA Oligos
E@sy Oligo®
Attention: The following specifications apply to the USA and Europe. All other countries, click here.
Simplify your life.
E@sy Oligo has been designed for simplicity. Order your primer at a fixed concentration and a fixed price. E@sy Oligo ships within 24 hours from order receipt, and is suitable for a wide range of applications including PCR and sequencing.
Sigma Life Science Logo
Genosys_button
Guaranteed yield
At least 3 OD (approx. 15 nmols*)
Format
100 µM concentration in de-ionized water
2 mL screw cap tubes
Length
15 to 30 mers
Quality
Fully deprotected and desalted
Bases
DNA (A, C, G or T)
Modifications
None
Backbone
Phosphodiester bond
Documentation
All Oligos are delivered with Quality Assurance document including yields, Tm (melting temperature), MW, and µg/OD
Quality control
Every oligo is quality checked by MALDI-TOF mass spectrometry
Maximum Order
48 oligos maximum per order for 24 hour shipment
Shipping Schedule
Ship within 24 hours of order receipt. Check with your local office for cut-off times.
Storage
See information on our oligos: storage recommendations
Price
Contact your local office for pricing and availability
*Estimate 1 OD = 5 nmols = 30 µg, for a 20 mer oligo