|
Simplify your life.
E@sy Oligo has been designed for simplicity. Order your primer at a fixed concentration and a fixed price. E@sy Oligo ships within 24 hours from order receipt, and is suitable for a wide range of applications including PCR and sequencing.
| Guaranteed yield |
At least 3 OD (approx. 15 nmols*) |
| Format |
100 µM concentration in de-ionized water 2 mL screw cap tubes |
| Length |
15 to 30 mers |
| Quality |
Fully deprotected and desalted |
| Bases |
DNA (A, C, G or T) |
| Modifications |
None |
| Backbone |
Phosphodiester bond |
| Documentation |
All Oligos are delivered with Quality Assurance document including yields, Tm (melting temperature), MW, and µg/OD |
| Quality control |
Every oligo is quality checked by MALDI-TOF mass spectrometry |
| Maximum Order |
48 oligos maximum per order for 24 hour shipment |
| Shipping Schedule |
Ship within 24 hours of order receipt. Check with your local office for cut-off times. |
| Storage |
See information on our oligos storage recommendations |
| Price |
Contact your local office for pricing and availability |
*Estimate 1 OD = 5 nmols = 30 µg, for a 20 mer oligo
|