Sigma-Aldrich
Life Science > Custom Oligos > Custom DNA Oligos > Product Lines > E@sy Oligo®
Custom DNA Oligos
E@sy Oligo®
Attention: Over the next few weeks, we will be updating our online order system and streamlining product specifications.
USA and Europe, Click Here
Simplify your life.
  • Shipped within 24 hours
  • Fixed price per oligo (15 to 30 mers)
Sigma_Proligo_Logo_SMALL
For unmodified DNA oligos, order E@sy Oligo® - designed to make your life easier. We guarantee to ship E@sy Oligo overnight, within 24 hours of receiving your order*. For your convenience, E@sy Oligo is available at a fixed price per oligo (15 to 30mers).
  • Guaranteed yield of at least 3 OD (approx. 15 nmols*)
  • Deprotected and desalted
  • Delivered in deionized water at a concentration of 100 µM
E@sy Oligo is suitable for a wide range of applications including PCR, sequencing and real-time quantitative PCR (as a primer).
*Please check local ordering cut-off times.
Specifications of Easy Oligo
Quality control
All of our oligos undergo vigorous process monitoring and strict quality control by MALDI-TOF mass spectrometry
Purification
Fully deprotected and desalted
Guarenteed yield
At least 3 OD (approx. 15 nmols*)
Length
15 to 30 mers
Bases
DNA (A, C, G or T)
Modifications
None
Backbone
Phosphodiester bond
Format
Delivered in de-ionized water at a concentration of 100 µM
Turn-around
time (TAT)
Shipped within 24 hours of receiving your order
Storage
and stability
Although oligonucleotides are stable in solution at 4°C for up to 2 weeks, Sigma-Proligo recommends storage should be at -20°C. Repetitive freeze-thaw cycles should be avoided by storing as aliquots. Storing at concentrations above 20 µM is recommended. Sigma-Proligo guarantees its oligonucleotides for 6 months, stored under the above conditions.
Shipment
Shipped by express delivery in solution in individual, transparent tubes
Oligonucleotide
Technical Data
Sheet
Oligonucleotides are delivered with an Oligonucleotide Technical Data Sheet, which includes oligo name, sequence, concentration, quantity in OD and nmols, Tm, MW, size, extinction coefficient and purification data.
Pricing
Please contact your local Sigma-Proligo representative
Ordering
Web orders only
*Estimate 1 OD = 5 nmols = 30 µg, for a 20 mer oligo
Literature available for download:
E@sy Oligo brochure
ORDERING
SERVICE & SUPPORT
CORPORATE
    Follow us on Twitter!  Join us on Facebook!  Join us on LinkedIn!  Watch our videos on YouTube!
Copyrights © 2010 Sigma-Aldrich Co. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.