|
Custom DNA Oligos
Fast Oligo™
|
|
When delivery speed is critical, order our Fast Oligo! We guarantee to ship Fast Oligo the same day as ordered. For your convenience, Fast Oligo is available at a fixed concentration and a fixed price.
Fast Oligo is suitable for a wide range of applications including PCR and sequencing.
| Guaranteed yield |
At least 3 OD (approx. 15 nmols*) |
| Format |
100 µM concentration in de-ionized water 2 mL screw cap tubes |
| Length |
15 to 30 mers |
| Quality |
Fully deprotected and desalted |
| Bases |
DNA (A, C, G or T) |
| Modifications |
None |
| Backbone |
Phosphodiester bond |
| Documentation |
All Oligos are delivered with Quality Assurance document including yields, Tm (melting temperature), MW, and µg/OD |
| Quality Control |
Every oligo is quality checked by MALDI-TOF mass spectrometry. |
| Maximum Order |
48 oligos maximum per order for next day delivery |
| Shipping Schedule |
Shipped same day. Check with your local office for cut-off times. |
Storage and Stability |
See information on our oligos: storage recommendations |
| Price |
Contact your local office for pricing and availability |
*Estimate 1 OD = 5 nmols = 30 µg, for a 20 mer oligo