|
Attention: Currently available in the USA and France.
When delivery speed is critical, order our Fast Oligo™! We guarantee to ship Fast Oligo the same day as ordered. For your convenience, Fast Oligo is available at a fixed concentration and a fixed price.
Fast Oligo is suitable for a wide range of applications including PCR and sequencing.
|
Guarenteed yield
|
At least 3 OD (approx. 15 nmols*)
|
|
Format
|
100 µM concentration in de-ionized water 2 mL screw cap tubes
|
|
Length
|
15 to 30 mers
|
|
Quality
|
Fully deprotected and desalted
|
|
Bases
|
DNA (A, C, G or T)
|
|
Modifications
|
None
|
|
Backbone
|
Phosphodiester bond
|
|
Documentation
|
All Oligos are delivered with Quality Assurance document including yields, Tm (melting temperature), MW, and µg/OD
|
|
Quality control
|
Every oligo is quality checked by MALDI-TOF mass spectrometry.
|
|
Maximum Order
|
48 oligos maximum per order for next day delivery
|
|
Shipping Schedule
|
Shipped same day. Check with your local office for cut-off times.
|
|
Storage and stability
|
See information on our oligos: storage recommendations
|
|
Price
|
Contact your local office for pricing and availability
|
*Estimate 1 OD = 5 nmols = 30 µg, for a 20 mer oligo
|