Long Oligo Specifications
| Yield |
Minimum 1 OD |
| Purification |
PAGE |
| Length |
Up to 250 bases |
| Backbone |
Phosphodiester / Phosphorothioate |
| Format |
Dry |
| Modifications |
Variety of modification options including Amines, Biotins, Fluorescent Dyes, and Phosphorylation |
| Packaging |
2 mL Tubes |
| Quality Controls |
Electrospray Ionization MS and OD by UV Spectroscopy |
| Technical Datasheet & Labels |
Includes yield, Tm (melting temperature), MW, and µg/OD
Duplicate set of labels with each order |
| Shipping Schedule |
12 business days |