Life Science > Custom Oligos > Custom DNA Oligos > Product Lines > One Price Primer
Custom DNA Oligos
One Price Primer
One Price Primers are supplied lyophilised in tubes.
  • Oligo length from 10 - 30 bases
  • Shipped within 24 - 48 hours
  • Deprotected and desalted
  • QC by statistical PAGE and MALDI-TOF mass spectrometry
  • QA documentation and additional vial label with every order
  • A, C, G, T bases only - no modifications
Sigma Life Science Logo
Genosys_button
For more information about One Price Primers, contact your local office