Life Science > Custom Oligos > Custom DNA Oligos > Product Lines > Ready-to-Use Primers
Custom DNA Oligos
Ready-to-Use Primers
Ready-to-Use oligos are normalized to 50 µM in 150 µL of TE. Format is suitable for applications such as PCR.
  • Oligo length from 10 - 30 bases
  • Normalized to 50 µM in 150 µL of TE
  • Shipped in a 2 mL tube at ambient temperature within 24 - 48 hours
  • Deprotected and desalted
  • QC by statistical PAGE and MALDI-TOF mass spectrometry
  • QA documentation and additional vial label with every order
Additional Specifications
  • A, C, G, T bases only - no modifications
  • TE Composition - 10 mM Tris HCL, 0.1 mM EDTA, pH 8.0
  • Maximum order size is 48 oligos per order
Sigma Life Science Logo
Genosys_button
For more information about Ready To Use Oligos, contact your local office