Life Science > Custom Oligos > Custom DNA Oligos > Product Lines > S-Oligos > RNA Oligonucleotide Synthesis (7-45 mers)
Custom DNA Oligos
RNA Oligonucleotide Synthesis (7-45 mers)
Attention: The following specifications apply to the USA and Europe.
All other countries, click here.
Genosys_button
Product Line:
Quality Desalted
Synthesis Scale 0.05 µmole 0.2 µmole 1.0 µmole
Standard RNA X X X
Mixed RNA/DNA/2-OMe/LNA® X X X
Phosphorothioate linkages (s-oligos) X X X
Guaranteed Yields (7-45 mers):
Synthesis Scale 0.05 µmole 0.2 µmole 1.0 µmole
Desalted 5 OD (150µg) 12 OD (360µg) 40 OD (1200µg)
RP-HPLC 1 OD (30µg) 2.5 OD (75µg) 13 OD (390µg)
PAGE 0.5 OD (15µg) 1 OD (30µg) 5 OD (150µg)
Purification:
Quality Purified
Synthesis Scale 0.05 µmole 0.2 µmole 1.0 µmole
RP-HPLC X X X
PAGE X X X
Quality
• fully deprotected and desalted, purified by PAGE or RP-HPLC on request
• it is highly recommended to use purified RNA for oligos > 30 mers.
Form
• RNA and 2' O-Methyl mixed oligos are sent dry
• Quantity (OD and nmoles) and concentration (µM and µg/µl)
Quality Controls
See information on our oligos: strict quality controls
Guarantee
Guaranteed sequence, quality and quantity
Please see yield table above for guaranteed minimum yields
Documentation
All Oligos are delivered with an Oligonucleotide Technical Data Sheet that can also be sent through e-mail
Additional Services
• Aliquoting
Synthesis Time
• 6 days for standard RNA
• 7 days for standard or PAGE purified mixed oligos
• 10 days for RP-HPLC purified oligos
Storage
Although oligonucleotides are stable at 4°C for up to 2 weeks, Sigma recommends storing at -20°C. Repetitive freeze-thaw cycles should be avoided by storing as aliquots. For long-term storage, RNA oligos should be dried.
LNA (Locked Nucleic Acids) is a registered trademark of Exiqon A/S.