Life Science > Custom Oligos > Custom DNA Oligos > Product Lines > Standard Synthesis
Custom DNA Oligos
Standard Synthesis
Sigma-Genosys offers a complete line of custom oligonucleotides for your research needs. Our proprietary platforms have the capacity to produce over 10,000 oligos per day in the U.S. and 5,000 per day at each of our international facilities.
Attention: Over the next few weeks, we will be updating our online order system and streamlining product specifications.
USA and Europe, Click Here
Sigma Life Science Logo
Genosys_button
 On this page:
Synthesis Scales
Our synthesis scales include 0.025 (3 O.D. Select), 0.05, 0.2, 1.0, 10, and 15µmole. (Not all available in all areas). Large scale synthesis is available upon request.
back to top
Purification
All Sigma-Genosys oligos include desalting and deprotection at no additional charge. For stringent applications, additional purification is available. Purification options include reverse-phase HPLC, PAGE, and reverse-phase cartridge (RP1). If you are unsure about the purification level you need for your research, see our technical bulletin entitled Selecting the Best Purification or contact our Technical Support staff for assistance.
back to top
Guaranteed Yield
Yield will vary based upon base composition, purification, oligo length, scale and modifications. That's why we have set minimum yield standards - guaranteed quantity. However, it is typical that you may receive more than the minimum yield guarantee. See our Yield Chart for details.
back to top
Shipping Schedule
Standard oligos will ship within 24 - 48 hours. Most modified oligos will ship within 5 days. See our Shipping Schedule for a complete list of expected shipment times. Our Customer Service department is available to answer any questions on your order status. And now - you can use our online order tracking to see first-hand where your oligo is within our process.
back to top
Guaranteed Quality
Oligos are routinely checked for length, purity, and composition. Our global quality control capabilities include MALDI-TOF mass spectrometry, electrospray ionization mass spectrometry (ESI-MS), capillary electrophoresis (CE), and polyacrylamide gel electrophoresis (PAGE). If you are dissatisfied and report an oligo problem within 30 days of order receipt, we will gladly remake your oligo free of charge or provide a credit for the purchase amount.
All orders are shipped with a Quality Assurance document that includes measured yields, melting temperature, MW and µg/OD with every oligo. Oligos in tubes are accompanied by an extra vial label for your records.
That's Not All
We have what the others don't - exceptional customer service and technical support. No other company gives you the hands-on care that Sigma-Genosys provides. From special price quotes to order confirmation to delivery updates, we will do what it takes to delight you with our service. Orders may be accepted by fax, e-mail or web.
back to top