Life Science > Custom Oligos > Custom DNA Oligos > Product Lines > Standard Synthesis
Custom DNA Oligos
Standard Synthesis
Standard Synthesis and Purification | Global Manufacturing | Our Commitment to Quality
Synthesis Scales • 0.025, 0.05, 0.2, 1.0, 10, and 15µmole
• mg - gram quantity manufacturing available upon request
Quality Fully deprotected and desalted, purified by RPC, HPLC or PAGE on request. See Oligonucleotide Purification Guidelines.
Form • Dried format, 2 mL screw cap tubes
• Concentration normalization available upon request
Quality Controls Every oligo is quality checked by MALDI-TOF mass spectrometry, electrospray ionization mass spectrometry (ESI-MS), capillary electrophoresis (CE), and/or polyacrylamide gel electrophoresis (PAGE).
Guarantee Guaranteed sequence, quality and quantity
(All quantities indicated are minimum guaranteed quantities delivered)
See our Yield Chart for details.
Documentation All Oligos are delivered with Quality Assurance document including yields, Tm (melting temperature), MW, and µg/OD. An extra vial label for your records accompanies oligos in tubes.
Additional Services • Concentration adjustment
• Aliquoting into tubes or well plates
• MSQC traces available upon request
Shipping Schedule • 24 hours for standard oligos in small quantity
• See Shipping Schedule for more details.
Global Manufacturing
Global operational facilities provide us with distinct advantages when compared to other manufacturers. We are able to provide a product with consistent global quality for projects that may require collaboration with scientists in other geographies. Sigma maintains disaster preparation plans and contingencies that mitigate circumstances arising from weather events such as floods, hurricanes, & power outages. With our global manufacturing capabilities and networking systems, we are able to continue the manufacturing process if our offices are threatened by natural disasters.
Our Commitment to Quality
Sigma has a controlled manufacturing process that includes: advanced scheduling tools using our production database (LIMS), redundant manufacturing equipment, dedicated order management team, 24hr/5 day manufacturing operation, and a world-class quality management system. All of our major manufacturing sites are ISO 9001:2000 or ISO14001:1996 certified.
As part of our quality management system, we require and measure continuous improvement through Plan-Do-Study-Act cycles and regularly review the results of these activities during our management reviews. Sigma employs an extensive array of performance metrics that are tracked, published, and reviewed at every level of our organization.
back to top
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Custom Oligos DNA