|
Custom Oligos
Custom iScale Oligos™
|
|
iScale Oligos are milligram and gram-quantity yields for in vivo basic research and in vitro high-throughput, research-tool, and diagnostic projects and kits.
| iScale Oligos DNA and RNA Specifications |
| Applies to every oligo |
Yields
- 50, 100, 250, 500, 750, and 1000 mg. Inquire for feasibility of gram-quantity synthesis
- Click the 'siRNA specifications' link below for yield information
|
Purification
- RP-HPLC, IE-HPLC, or both to ≥85% depending on application
|
Quality Control
- Analytical IE-HPLC and MALDI-TOF or ESI mass spectrometry
|
Final Form
- Powder in sterile, nuclease-free, pyrogen-free tubes or bottles
|
Documentation
- Certificate of analysis including HPLC and mass spec traces
|
Guarantee
- Sequence, purity, and quantity up to 50 DNA bases
- Sequence, purity, and quantity up to 35 RNA bases
|
Shipping
- 2 weeks for 1000 mg or less
|
| iScale Oligos DNA and RNA Extras |
| Additional fees apply |
Modifications
- Most standard mods including phosphorothioate oligos are available
|
In vivo Options
- Filtration, dialysis, conductivity measurements, and endotoxin testing
|
iScale Oligos are manufactured in an ISO 9001:2008 certified facility.