Life Science > Custom Oligos > Custom iScale Oligos™ > In Vivo Purification and Quality Control
Custom Oligos
In Vivo Purification and Quality Control
To ensure that oligos are tolerated by host animals, Sigma® offers several additional options with iScale Oligos™ DNA and RNA.
iScale Oligos DNA and RNA Extras Explained
Recommended for animal work
Filtration
  • Passage through a 0.22 µm membrane minimizes bacteria, molds, and other contaminants
Dialysis
  • Toxic synthesis, cleavage, deprotection, and purification solvents and salts are exchanged for those that are physiologically acceptable
Conductivity Measurements
  • Dialysis ensures that salts are in the range of 10 – 100 µS m-1 at a 50 µM concentration
Endotoxin Testing
  • 1 EU mg-1 is the upper limit allowed (values are typically in the range of 0.1 – 1 EU mg-1)
  • Endotoxin testing (LAL assay) detects LPS, a constituent molecule found in the outer membrane of gram negative bacteria
  • LPS must be below an acceptable threshold, or the host will die from toxic shock
back to Custom iScale Oligos
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Custom Oligos DNA