Peptide Synthesis
Custom Peptide Library | PEPscreen®
Reveal Your Target of Interest
Experience affordable and fast delivery of custom peptide libraries for target identification. The PEPscreen peptide library is configured in a convenient plate format, perfect for screening applications, epitope mapping and peptide microarray production.
PEPscreen Webinar
Watch the webinar and learn about the importance of utilizing custom peptide libraries for different types of screening applications. You will learn about designing the optimal peptide libraries as well as epitope mapping and ELISPOT assays involving peptide libraries. You will also learn from other researchers how they are utilizing peptide libraries in their specific applications.
Design
Design your library with ease using our convenient design tool. It enables you to design overlapping peptide fragment libraries, alanine scanning libraries, truncation libraries, and positional scanning libraries.
You’ll be able to:
  • Set custom sequence parameters (peptide length and amino acid offset)
  • Format the sequence output
  • Export the custom peptide library sequences (MS Word, Excel, etc.)
Benefits
We have developed a proprietary custom peptide synthesis platform utilizing Fmoc chemistry, providing a solution to enhance peptide screening applications. Using this technology we can provide high-quality peptide libraries with:
  • Low variability: Completely automated synthesis eliminates errors
  • Fast delivery: Receive your library in less than 7 business days!
  • Unsurpassed flexibility: Include multiple peptide lengths and designs in a single library
  • Easy-to-use formatting: Individual tubes are in 96-well format making it compatible with high-throughput robotic assays and eliminates cross-contamination. Individual tubes are triple-labeled for maximum user flexibility
Product Specifications
  • Amount: 0.5 – 2mg; 2mg – 5mg
  • Purity level: unpurified
  • Sequence length: 6 – 20mers
  • Modifications available
  • QC: MALDI-MS on 100% of peptides
  • Format: supplied dried in 96-well tube rack
  • Manufacturing Time: 4 – 7 business days
  • Minimum order size: 24 peptides
Ordering Information
To request a quotation or place your order, download the excel template below and follow the instructions.
Learn More
Our PEPscreen brochure offers in depth information from designing a peptide library to sequence optimization. It is available for download (pdf) or you can browse the web version.
Here are additional technical resources:
Satisfied Customers
Our customers have shared with us some of the ways they’ve used PEPscreen to further their research. Read all the application data they have generated. Have you used PEPscreen and would like to share your data?
Need More Info?
We are happy to provide additional information that is specific to your research. Email us at peptides@sial.com to request additional information.