Sigma-Aldrich
Life Science > Custom Oligos > Peptide Synthesis > Product Lines > PEPscreen® Peptides
Peptide Synthesis
PEPscreen® Peptides
Sigma® has developed a proprietary custom peptide synthesis platform utilizing Fmoc chemistry, providing a solution to enhance peptide screening applications. Large numbers of peptides, in 96-well arrays, are delivered quickly and affordably. MALDI-TOF mass spec is performed on 100% of the samples.
  • Product Specifications
  • Internal Testing of Synthesis Platform
  • Technical Resources
  • Application Data
  • Share your expertise
PEPscreen<sup>®</sup> calculator
PEPscreen Library
Design Tool
PEPscreen<sup>®</sup> – An Enabling Technology for Peptide-Based Drug Discovery
PEPscreen Brochure Online
Product Specifications:
  • Low cost per peptide
  • Amount: 0.5 - 2 mg or 2 - 5 mg
  • Length: 2 - 20 amino acids
  • Comprehensive Modification Offering
  • MALDI-ToF Mass Spectrometry on 100% of peptides
  • Peptides supplied dried and in 96-well tube rack format
  • Dispatch: approximately 7 business days
  • Minimum order size is 24 peptides
Internal Testing of Synthesis Platform:
  • We routinely run tests of this platform to ensure the quality of the product.
  • Average Crude Purity of 10mers: ~86%
  • Average Crude Purity of 15mers: ~73%
  • Average Crude Purity of 20mers: ~61%
  • Purity of ACP peptide at different positions on a 96-well plate: ~92% +/- 2%
Technical Resources:
  • Epitope Mapping and Vaccine Research: Vaccination as a Strategic Medical Option
  • Frequently Asked Questions for PEPscreen Peptides
  • Generalized Steps of the ELISPOT Assay Procedure
  • Guidelines for Dissolving PEPscreen Peptide Sets (253 Kb PDF)
  • Storage and Handling Guidelines (689 Kb PDF)
Application Data Now Available!
The following application data was received from customers using our PEPscreen Technology:
  • Diverse CD4 Response to Vaccinia Virus in Human is Revealed by Proteome-Wide T cell Profiling
  • Binding and Kinetic Analysis of Protein Interactions Probed by Label-Free SpotMatrix SPR Technology in a Peptide Chip Format
  • Cross-CladeCD8+ T-Cell Responses with a Preference for the Predominant Circulating Clade
  • Development of Peptide-based Microarrays for Screening Antibodies from Patients with Autoimmune Diseases
  • High-throughput Screening of Peptide Substrates for Tyrosine Kinases Using PEPscreen Custom Peptide Library
  • Use of Peptide Libraries to Investigate the Evolutionary Preservation of Antimicrobial Properties of Complement Peptide C3a from Invertebrates to Humans
  • Use of PEPscreen Libraries to Identify CD8 Epitopes Presented During Malaria Infection
  • Use of PEPscreen Peptides to Investigate CTL Class I MHC and Th Class II Responses to AAV-hF.IX Gene Therapy
  • A Quantitative Mass Spectrometry-Based Approach for Identifying Protein Kinase Clients and Quantifying Kinase Activity
  • Use of PEPscreen Peptides to Investigate Protein Farnesyltransferase Substrate Specificity