|
Sigma-Genosys
Custom RNA Oligos
|
Attention: The following specifications apply to the USA and Europe. All other countries, click here.
|
Product Line:
| Quality |
Desalted |
| Synthesis Scale |
0.05 µmole |
0.2 µmole |
1.0 µmole |
| Standard RNA |
X |
X |
X |
| Mixed RNA/DNA/2-OMe/LNA® |
X |
X |
X |
| Phosphorothioate linkages (s-oligos) |
X |
X |
X |
Guaranteed Yields (7-45 mers):
| Synthesis Scale |
0.05 µmole |
0.2 µmole |
1.0 µmole |
| Desalted |
5 OD (150µg) |
12 OD (360µg) |
40 OD (1200µg) |
| RP-HPLC |
1 OD (30µg) |
2.5 OD (75µg) |
13 OD (390µg) |
| PAGE |
0.5 OD (15µg) |
1 OD (30µg) |
5 OD (150µg) |
Purification:
| Quality |
Purified |
| Synthesis Scale |
0.05 µmole |
0.2 µmole |
1.0 µmole |
| RP-HPLC |
X |
X |
X |
| PAGE |
X |
X |
X |
|
Quality
|
• fully deprotected and desalted, purified by PAGE or RP-HPLC on request • it is highly recommended to use purified RNA for oligos > 30 mers.
|
|
Form
|
• RNA and 2' O-Methyl mixed oligos are sent dry • Quantity (OD and nmoles) and concentration (µM and µg/µl)
|
|
Quality Controls
|
See information on our oligos: strict quality controls
|
|
Guarantee
|
Guaranteed sequence, quality and quantity Please see yield table above for guaranteed minimum yields
|
|
Documentation
|
All Oligos are delivered with an Oligonucleotide Technical Data Sheet that can also be sent through e-mail
|
|
Additional Services
|
• Aliquoting
|
|
Synthesis Time
|
• 6 days for standard RNA • 7 days for standard or PAGE purified mixed oligos • 10 days for RP-HPLC purified oligos
|
|
Storage
|
Although oligonucleotides are stable at 4°C for up to 2 weeks, Sigma recommends storing at -20°C. Repetitive freeze-thaw cycles should be avoided by storing as aliquots. For long-term storage, RNA oligos should be dried.
|
LNA (Locked Nucleic Acids) is a registered trademark of Exiqon A/S.