Sigma-Aldrich
Life Science > Custom Oligos > Custom RNA Oligos
Custom Oligos
Custom RNA Oligos
Product Line
Quality Desalted
Synthesis Scale 0.05 µmole 0.2 µmole 1.0 µmole
Standard RNA X X X
Mixed RNA/DNA/2-OMe/LNA® X X X
Phosphorothioate linkages (s-oligos) X X X
Guaranteed Yields (7-45 mers)
Synthesis Scale 0.05 µmole 0.2 µmole 1.0 µmole
Desalted 5 OD (150µg) 12 OD (360µg) 40 OD (1200µg)
RP-HPLC 1 OD (30µg) 2.5 OD (75µg) 13 OD (390µg)
PAGE 0.5 OD (15µg) 1 OD (30µg) 5 OD (150µg)
Purification
Quality Purified
Synthesis Scale 0.05 µmole 0.2 µmole 1.0 µmole
RP-HPLC X X X
PAGE X X X
Quality • fully deprotected and desalted, purified by PAGE or RP-HPLC on request
• it is highly recommended to use purified RNA for oligos > 30 mers.
Form • RNA and 2' O-Methyl mixed oligos are sent dry
• Quantity (OD and nmoles) and concentration (µM and µg/µl)
Quality Controls Every oligo is quality checked using one or more of the following techniques:  MALDI-TOF mass spectrometry, electrospray ionization mass spectrometry (ESI-MS), capillary electrophoresis (CE), or polyacrylamide gel electrophoresis (PAGE).
Guarantee Guaranteed sequence, quality and quantity
Please see yield table above for guaranteed minimum yields
Documentation All oligos are delivered with a Quality Assurance document including yields, Tm (melting temperature), MW, and µg/OD.
Additional Services • Aliquoting
Synthesis Time • 6 days for standard RNA
• 7 days for standard or PAGE purified mixed oligos
• 10 days for RP-HPLC purified oligos
Storage Although oligonucleotides are stable at 4°C for up to 2 weeks, Sigma recommends storing at -20°C. Repetitive freeze-thaw cycles should be avoided by storing as aliquots. For long-term storage, RNA oligos should be dried.
LNA (Locked Nucleic Acids) is a registered trademark of Exiqon A/S.