Life Science > Custom Oligos > Custom RNAtural Oligos
Custom Oligos
Custom RNAtural Oligos
Only in recent years have in vitro transcription and chemical synthesis made it worthwhile for scientists to consider directly working with the intractable RNA molecule. Yet, both technologies have limitations. In vitro transcription tends to be useful for long sequences and chemical synthesis for abundant quantities. Combined, they would make a novel and ideal technology.
In partnership with MitoProd, Sigma is pleased to announce that this novel and ideal technology now exists. Introducing RNAtural—RNA, The Natural Way. RNAtural is biologically-produced, custom RNA sequences manufactured up to thousands of bases in milligram yields. Under ISO 9001 conditions, yeast are used to produce the desired sequence as shown in the following animation:
Click to view the Custom RNAtural Animation in a New Window
Click to open the animation in a new window
All that is needed is a sequence — we will do the rest. Sigma and MitoProd want to partner with you for all of your research, pre-clinical, and diagnostic RNA sequence needs.
Custom RNAtural – RNA Request Form
Request Pricing and Feasibility for RNA Sequence
RNAtural Specifications
Sigma Life Science Logo
Capacity 1 native, single-stranded RNA sequence per order
Lengths 30 – 5,000 bases
Yields 10 – 1,000 mg
Purification Options HPLC-IE >80% (please inquire for higher homogeneities)
Available Quality Control Length measurements, purity measurements, RNase testing, and endotoxin testing
Format Sterile tubes guaranteed to be RNase, DNase, and pyrogen free
Documentation Each sequence is delivered with a certificate of analysis
Delivery Please inquire
Storage Short-term and long-term protocols provided
Pricing Please inquire
Ordering Online, email, or fax
Manufactured under ISO 9001 conditions.
back to top