Life Science > Custom Oligos > Custom DNA Probes
Custom Oligos
Custom DNA Probes
Bioconfident. Unleash superior assay performance with Sigma's custom qPCR probes
Whether your application is genotyping, gene expression, pathogen detection, or other, Sigma's wide selection of reporter and quencher dyes for quantitative PCR (qPCR) probes will support all real time and end-point detection chemistries.
Dual-Labeled Probes: Fluorescent probes for Real-Time Quantitative PCR and other Applications.
Molecular Beacons: Structured probes that are highly sensitive, sequence-specific and used for sequence detection in real-time qPCR and in vitro studies.
Scorpions™ Probes: Highly sensitive, sequence-specific, bi-labeled fluorescent probe/primer hybrids designed for real-time qPCR.
LightCycler® Probes: Highly sensitive and sequence-specific fluorescent probes designed for use with the Roche™ LightCycler instruments.
Product Highlights
qPCR and MIQE Seminar Series
OligoArchitect™ Primer and Probe Design Solutions
Choosing the Right Probe Type
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Custom Oligos DNA
OligoArchitect Online v2.0
Extract-N-Amp: Sample to PCR in under 15 minutes.
CleanAmp dNTPs