Life Science > Custom Oligos > Custom DNA Probes > Product Lines > Choosing the Right Probe
Custom DNA Probes
Choosing the Right Probe
There are a selection of possible probes that can be used for qPCR and each has advantages for different applications. The summary below highlights the applications of commonly used probes.
Application Reference Guide
(For additional product information, click on the column headers below.)
  SYBR® Green I Dual-Labeled Probes Dual-Labeled LNA® Probes Molecular Beacons* LightCycler® Probes* Scorpions™ Probes*
APPLICATION
Mass screening • •          
Microarray validation • •        
Multiple target genes / few samples        
SNP detection     • • • •
Allelic discrimination     • • • •
Pathogen detection • • • •
Multiplex   • • • • • • • •
Viral load quantification   • • • •
Gene expression • • • • • • • • • •
Gene copy determination   • • • •
End point genotyping       • •   • •
End point genotyping     • •    
*LNA can be incorporated into the probes for improved specificity.
back to top
Spectral Properties Table
Dye Max.
EX (nm)
Max.
EM (nm)
Compatible
Quencher
6-FAM™ 494 515 BHQ-1, TAMRA
JOE™ 520 548 BHQ-1, TAMRA
TET™ 521 536 BHQ-1, TAMRA
Cal Fluor® Gold 5401 520 548 BHQ-1
HEX™2 535 555 BHQ-1, TAMRA
Cal Fluor® Orange 5602 522 541 BHQ-1
TAMRA™ 555 576 BHQ-2
Cy3® 550 570 BHQ-2
Quasar® 5703 548 566 BHQ-2
Cal Fluor® Red 5904 565 588 BHQ-2
ROX™ 573 602 BHQ-2
Texas Red® 583 603 BHQ-2
Cy5® 651 674 BHQ-3
Quasar 6705 647 667 BHQ-3
Cy5.5® 675 694 BHQ-3
1JOE/TET alternative
2VIC® alternative
3Cy3 alternative
4TAMRA alternative
5Cy5 alternative
back to top
Fluorophores and Instrument Compatibility Table
Platform SYBR
Green I
FAM HEX JOE ROX TET Cy3 Cy5 TAMRA Texas
Red
LC Red
640
LC Red
705
ABI 7900HT          
ABI 7300            
ABI 7500      
ABI 7700            
ABI 7000              
Bio-Rad iCycler iQ
Bio-Rad Opticon® 2              
Bio-Rad Chromo4™    
Stratagene Mx4000®    
Stratagene Mx3000P®    
Stratagene Mx3005P®    
Roche LightCycler®                
Roche LightCycler 2              
Roche LightCycler 480            
Cepheid SmartCycler®              
Cepheid SmartCycler II            
Corbett Rotor-Gene™ 6000      
Eppendorf Mastercycler® realplex          
Note: Not all qPCR instruments or fluorophores are listed. Contact the instrument manufacturer for details on compatible fluorophores
Sigma® is a licensed supplier of a variety of dyes and quenchers and continually adds to its portfolio of new chemistries. For assistance in the design of your probe and/or assays, submit your request at sigma.com/designmyprobe.
back to top
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Custom Oligos DNA