Sigma-Aldrich
Life Science > Custom Oligos > Custom DNA Probes > Product Lines > Dual-Labeled Probes
Custom DNA Probes
Dual-Labeled Probes
Whether your application is genotyping, gene expression, pathogen detection, or other, Sigma's wide selection of reporter and quencher dyes for quantitative PCR (qPCR) probes will support all real time and end-point detection chemistries.
  • When to Choose Dual-Labeled Probes
  • Benefits of Using Dual-Labeled Probes
  • Add LNA® to Your Probe
  • How Dual-Labeled Probes Work
  • Product Features
  • Guaranteed Yields
  • Spectral Properties Table
Choose Dual-Labeled Probes for:
  • Gene expression
  • Multiplex assay development
  • Pathogen detection
  • Viral load quantitation
  • Microarray validation
  • si/shRNA knockdown
  • Gene copy determination
  • Mutation detection
  • Allelic discrimination
  • SNP detection
Dual-labeled probes are highly sensitive, bi-labeled fluorescent probes and are designed to be sequence specific. They can be used with most real-time quantitative PCR instruments and multiplex analysis systems due to their straightforward design and the extensive range of fluorophores.
Benefits of Using Dual-Labeled Probes include:
  • Design simplicity for sequence specificity
  • Increased sensitivity
  • Extensive availability of fluorophore combinations
Add LNA to Your Probe for:
  • Increased thermal stability and hybridization specificity
  • Greater accuracy in gene quantitation and allelic discrimination
  • Easier and more sensitive probe designs for problematic target sequences
back to top
How Dual-Labeled Probes Work
A dual-labeled probe is a single-stranded oligonucleotide labeled with two different dyes. A reporter dye is located at the 5' end and a quencher molecule located at the 3' end. The quencher molecule inhibits the natural fluorescence emission of the reporter dye by Forster-type energy transfer. The illustration below depicts the mechanism.
How Dual-Labeled Probes Work Image
The primer is elongated by the polymerase and the probe binds to the specific DNA template, hydrolysis releases the reporter dye from the probe/target hybrid, causing an increase of fluorescence. The measured fluorescence signal is directly proportional to the amount of target DNA.
Product Features include:
  • Available in lengths of 15–40 bases
  • Simple pricing – HPLC purification and base charges are included
  • Stringent quality control including electrospray mass spectroscopy
  • Shipped within 3–5 business days for 6-FAM™, HEX™, or TET™ labeled
  • Shipped in an amber tube to protect dye integrity
  • Custom formats available (normalization, special plates, etc.) Sigma's probes are provided in a format to simplify your experimental planning
back to top
Guaranteed Yields
Guaranteed
OD Yield
Approx. No.
of nmoles
Approx. No.
of µg
Approx. No.
of Reactions*
1 4 32 800
3 12 96 2,400
5 20 160 4,000
10 40 320 8,000
*Estimate is based on 4 nmoles or 32 µg for 1 OD and 200 nM in 25 µl reaction (5.0 pmol/reaction). Estimate is based on an average sequence length of 25 bases.
The most common fluorophore and quencher combinations are listed below:
Spectral Properties Table
Dye Max.
EX (nm)
Max.
EM (nm)
Compatible
Quencher
6-FAM™ 494 515 BHQ-1, TAMRA
JOE™ 520 548 BHQ-1, TAMRA
TET 521 536 BHQ-1, TAMRA
Cal Fluor® Gold 5401 522 541 BHQ-1
HEX2 535 555 BHQ-1, TAMRA
Cal Fluor® Orange 5602 540 561 BHQ-1
TAMRA™ 555 576 BHQ-2
Cy3® 550 570 BHQ-2
Quasar® 5703 548 566 BHQ-2
Cal Fluor® Red 5904 565 588 BHQ-2
ROX™ 573 602 BHQ-2
Texas Red® 583 603 BHQ-2
Cy5® 651 674 BHQ-3
Quasar 6705 647 667 BHQ-3
Cy5.5® 675 694 BHQ-3
1JOE/TET alternative
2VIC® alternative
3Cy3 alternative
4TAMRA alternative
5Cy5 alternative
Label License Statement
Sigma® is a licensed supplier of a variety of dyes and quenchers and continually adds to its portfolio of new chemistries. For assistance in the design of your dual-labeled probes and assays, submit your request at sigma.com/designmyprobe.