Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Dual-Labeled DNA Probes
Reporters Quenchers
Spectral properties of fluorophores
The table below details the most commonly used dyes for the fluorescent probes, together with their excitation and emission data. Please enquire for any other labels.
Quenchers
Maximum absorbance (nm)
Maximum emission (nm)
BHQ-1 Dark Quencher
535
None
BHQ-2 Dark Quencher
579
None
DABCYL Dark Quencher
453
None
DABCYL-dT Dark Quencher
453
None
TAMRA
550
576
Labels
Maximum absorbance (nm)
Maximum emission (nm)
6-FAM
495
517
Cy3
550
570
Cy5
650
667
Cy5.5
675
694
Fluorescein
495
520
HEX
537
553
JOE
520
548
LightCycler Red 610
590
610
LightCycler Red 640
625
640
LightCycler Red 670
650
670
LightCycler Red 705
680
705
Oregon Green 488
495
521
Oregon Green 500
499
519
Oregon Green 514
506
526
Rhodamine
564
603
6-CarboxyRhodamine 6G
524
557
Rhodamine Green
504
532
Rhodamine Red
570
590
ROX
581
607
TAMRA
550
576
TET
521
538
Texas Red
589
610