Dual-Labeled DNA Probes
LNA Oligos
Attention: The following specifications apply to the USA and Europe. All other countries, click here.
Speed-up your discovery.
  • Superior hybridization characteristics
  • Enhanced biostability
Sigma Life Science Logo
Genosys_button
LNA Bibliography
What is LNA?
LNA is a novel type of nucleic acid analog that contains a
2'-O, 4'-C methylene bridge. This bridge restricts the flexibility of the ribofuranose ring and locks the structure into a rigid C3-endo conformation, conferring enhanced hybridization performance and exceptional biostability.
PP_LNA_Image1
LNA® oligos are innovative oligonucleotides that contain Locked Nucleic Acids™ (LNA) to provide superior hybridization characteristics and enhanced biostability compared to conventional DNA oligonucleotides.
LNA-containing oligos are provided as follows:
  • Deprotected and desalted
  • Purified by PAGE, RPC or RP-HPLC upon request
  • Delivered in solution in deionized water or dried down
  • Systematic QC by MALDI-TOF mass spectrometry analysis and - depending upon the nature of the oligonucleotide requested - by RP-HPLC or IE-HPLC
  • Please contact Sigma-Genosys for expected TATs
LNA oligos are composed of either 100% Locked Nucleic Acid (LNA) bases or a mixture of LNA and DNA bases. However, only oligonucleotides that are between 7 and 15 mers in length can be made up of 100% LNA. Oligos heavy in LNA content may exhibit problematic self-complementarity. Therefore, for oligonucleotides longer than 15 mers, we recommend decreasing the LNA content as the oligonucleotide increases in length, as follows:
Length
of LNA oligo
7 - 15 mers
16 - 25 mers
26 - 40 mers
41 - 60 mers
Percentage
of LNA
up to 100%
up to 60%
up to 40%
up to 20%
Please contact Sigma-Genosys design specialists to discuss application-specific LNA design.
LNA oligos are available at our standard synthesis scales (0.05, 0.2, 1 µmole). LNA bases may also be incorporated into our Real-time qPCR probes. Consult with your local office for guaranteed yield and delivery times.
LNA oligonucleotides can be designed to enhance a wide variety of genomic applications and technologies that rely upon the use of oligonucleotides:
  • Hybridization probes for the detection of nucleic acids and genotyping
  • Padlock probes, modified Taqman™ probes and probes for in situ hybridization
  • Capture probes for target enrichment, sample preparation and affinity chromatography
  • Allele-specific PCR1
  • Antisense oligonucleotide technology
  • Oligonucleotides for therapeutic or diagnostic applications, such as, aptamers, ribozymes, decoy oligonucleotides and siRNA
  • PCR-based applications
In most respects, LNA oligonucleotides can be used in the same manner as DNA oligonucleotides: it is soluble in aqueous buffers, can be ethanol precipitated, dried and resuspended, and analyzed by gel electrophoresis, HPLC and MALDI-TOF.
Specifications of LNA Oligos
Quality control
Every oligo is quality checked by MALDI-TOF mass spectrometry, electrospray ionization mass spectrometry (ESI-MS), capillary electrophoresis (CE), and/or polyacrylamide gel electrophoresis (PAGE).
Purification
Fully deprotected and desalted
Purified by PAGE, RPC or RP-HPLC upon request
Length
7 to 15 mers: up to 100% LNA base content
7 to 60 mers: LNA / DNA mix
Bases
LNA (+A, +C, +G or +T) (+ denotes the LNA base)
Backbone
Phosphodiester and/or phosphorothioate bonds
Form
Dried format, 2 mL screw cap tubes. Concentration normalization available upon request.
Storage and stability
See information on our oligos: storage recommendations
Shipment
Shipped by express delivery, in individual tubes
Documentation
All Oligos are delivered with Quality Assurance document including yields, Tm (melting temperature), MW, and µg/OD. An extra vial label for your records accompanies oligos in tubes.
Services available upon request
• Aliquoting
• Adjusted concentration
• Design support
• Additional services may increase turn-around time
• Plate services
Pricing
Please contact your local Sigma-Genosys representative
1 PCR (Polymerase Chain Reaction) is patented by Hoffman La Roche Ltd.