Dual-Labeled DNA Probes
LNA Oligos
Attention: Over the next few weeks, we will be updating our online order system and streamlining product specifications.
USA and Europe, Click Here
Speed-up your discovery.
  • Superior hybridization characteristics
  • Enhanced biostability
Sigma_Proligo_Logo_SMALL
LNA Bibliography
What is LNA?
LNA is a novel type of nucleic acid analog that contains a
2'-O, 4'-C methylene bridge. This bridge restricts the flexibility of the ribofuranose ring and locks the structure into a rigid C3-endo conformation, conferring enhanced hybridization performance and exceptional biostability.
PP_LNA_Image1
LNA® oligos are innovative oligonucleotides that contain Locked Nucleic Acids™ (LNA) to provide superior hybridization characteristics and enhanced biostability compared to conventional DNA oligonucleotides.
LNA-containing oligos are provided as follows:
  • Deprotected and desalted
  • Purified by PAGE, RPC or RP-HPLC upon request
  • Delivered in solution in deionized water or dried down
  • Systematic QC by MALDI-TOF mass spectrometry analysis and - depending upon the nature of the oligonucleotide requested - by RP-HPLC or IE-HPLC
  • Please contact Sigma-Proligo for expected TATs
LNA oligos are composed of either 100% Locked Nucleic Acid (LNA) bases or a mixture of LNA and DNA bases. However, only oligonucleotides that are between 7 and 15 mers in length can be made up of 100% LNA. Oligos heavy in LNA content may exhibit problematic self-complementarity. Therefore, for oligonucleotides longer than 15 mers, we recommend decreasing the LNA content as the oligonucleotide increases in length, as follows:
Length
of LNA oligo
7 - 15 mers
16 - 25 mers
26 - 40 mers
41 - 60 mers
Percentage
of LNA
up to 100%
up to 60%
up to 40%
up to 20%
Please contact Sigma-Proligo design specialists to discuss application-specific LNA design.
Guaranteed Yields, Labels, Modifications and Turn-Around Time for LNA oligos
PP_LNA_Image2
LNA oligonucleotides can be designed to enhance a wide variety of genomic applications and technologies that rely upon the use of oligonucleotides:
  • Hybridization probes for the detection of nucleic acids and genotyping
  • Padlock probes, modified Taqman™ probes and probes for in situ hybridization
  • Capture probes for target enrichment, sample preparation and affinity chromatography
  • Allele-specific PCR1
  • Antisense oligonucleotide technology
  • Oligonucleotides for therapeutic or diagnostic applications, such as, aptamers, ribozymes, decoy oligonucleotides and siRNA
  • PCR-based applications
In most respects, LNA oligonucleotides can be used in the same manner as DNA oligonucleotides: it is soluble in aqueous buffers, can be ethanol precipitated, dried and resuspended, and analyzed by gel electrophoresis, HPLC and MALDI-TOF.
Specifications of LNA Oligos
Quality control
All of our oligonulceotides undergo vigorous process monitoring and strict quality control. Length and labeling are systematically controlled by PAGE or mass spectrometry analysis and - depending on the nature of the oligonucleotide requested, by RP-HPLC or IE-HPLC. Quantity is determined by UV absorbance at 260 nm.
Purification
Fully deprotected and desalted
Purified by PAGE, RPC or RP-HPLC upon request
Length
7 to 15 mers: up to 100% LNA base content
7 to 60 mers: LNA / DNA mix
Bases
LNA (+A, +C, +G or +T) (+ denotes the LNA base)
DNA (A, C, G or T), inosine, degenerate bases (wobbles)
Backbone
Phosphodiester and/or phosphorothioate bonds
Format
Delivered in solution in deionized water or dried down, upon request
Storage and stability
Although oligonucleotides are stable in solution at 4°C for up to 2 weeks, Sigma-Proligo recommends storage should be at -20°C. Repetitive freeze-thaw cycles should be avoided by storing as aliquots. Storing at concentrations above 20 µM is recommended. Oligonucleotides with fluorescent labels should be protected from light. Sigma-Proligo guarantees its oligonucleotides for six months, stored under the above conditions.
Shipment
Shipped by express delivery, in individual tubes
Oligonucleotide
Technical Data
Sheet
Oligonucleotides are delivered with an Oligonucleotide Technical Data Sheet, which includes oligonucleotide name, sequence, concentration, quantity in OD and nmols, Tm , MW, size, extinction coefficient and purification information.
Services available upon request
• Aliquoting
• Adjusted concentration
• Design support
• Additional services may increase turn-around time
• Plate services
Pricing
Please contact your local Sigma-Proligo representative
Ordering
On-line at www.proligo.com, by email or by fax
1 PCR (Polymerase Chain Reaction) is patented by Hoffman La Roche Ltd.
Literature available for download:
LNA Oligos Speed-up Discovery (136 K PDF)
LNA Fluorescent Probes for RT QPCR (271 K PDF)