Sigma-Aldrich
Life Science > Custom Oligos > Custom DNA Probes > Product Lines > LNA Probes > Black Hole Quenchers
Dual-Labeled DNA Probes
Black Hole Quenchers
Some commonly used quenchers-such as DABCYL and TAMRA-suffer from a number of drawbacks, including poor spectral overlap between the fluorescent dye and quencher molecule (DABCYL) or inherent fluorescence of the quencher (TAMRA)-resulting in a relatively poor signal-to-noise ratio. Black Hole Quencher™ (BHQ™) molecules have been developed to overcome these drawbacks.
Benefits:
  • No native fluorescence, resulting in lower background fluorescence
  • Increases signal-to-noise ratios, providing higher sensitivity
  • Accesses the visible spectrum into near-IR for reporting (480 to 730 nm)
  • Maximizes spectral overlap, increasing efficiency of quenching
  • Enables wider choice of reporter dyes for multiplexing assays
Absorption Spectra of BHQ dyes among the most commonly used dyes.
Black_Hole_Image1
Black_Hole_Image2