Life Science > Custom Oligos > Custom DNA Probes > Product Lines > LNA Probes > LNA® Dual-Labeled Fluorogenic Probes & Molecular Beacons
Dual-Labeled DNA Probes
LNA® Dual-Labeled Fluorogenic Probes & Molecular Beacons
Attention: The following specifications apply to the USA and Europe. All other countries, click here.
For your convenience, LNA dual-labeled fluorogenic probes and LNA molecular beacons are available at a fixed price per oligonucleotide, within the following parameters:
Sigma Life Science Logo
Genosys_button
  • Up to 8 LNA bases
  • Deprotected, desalted and purified by PAGE or RP-HPLC
  • Available in lengths of 7 to 40 mers
  • Delivered dried in individual, opaque tubes
  • Shipped in 7 working days of receiving your order, pending successful QC validation*
Guaranteed yields of LNA dual-labeled fluorogenic probes and
LNA molecular beacons
Guaranteed yield (OD)
Approximate yield (nmols**)
Estimated assays (reactions***)
1
5
500
3
15
1,500
5
25
2,500
10
50
10,000
** Estimate 1 OD = 5 nmols = 30 µg, for a 20 mer oligo
*** Estimated number of assays based on a 50 µl reaction volume.
Please enquire for alternative quantities.
Labels for LNA dual-labeled fluorogenic probes and
LNA molecular beacons
5'-end reporter
3'-end quencher
6-FAM™, HEX™ or TET™
TAMRA™, DABCYL, BHQ™-1
or BHQ-2
Please inquire about optional dye labels.
Figure 1: LNA dual-labeled fluorogenic probes vs. TaqMan® MGB probes in 5' nuclease PCR assays. (Amplification of Staphylococcal enterotoxin gene in a Staphylococcus aureus strain11).
PP_LNAFP_QPCR
Key
PP_LNAFP_Key
LNA dual-labeled fluorogenic probe
PP_LNAFP_Key_Green
TaqMan MGB probe
OD = optical density
* Please check with your Sigma-Genosys representative for local delivery times.
Specifications are subject to change.
redcube1
qPCR Total Solution Seminar Series
Seminars begin February 7th, Register Today
Image of Beacon Designer: Software for Real Time PCR Oligo Design
LNA—Notice of limited rights: Sigma, Sigma-Life Science are only licensed to sell Dual Labeled Fluorescent Probes, LightCycler Probes, Scorpion Probes and Molecular Beacons containing LNA (Locked Nucleic Acids) for use in Real-Time PCR.