|
For your convenience, LNA dual-labeled fluorogenic probes and LNA molecular beacons are available at a fixed price per oligonucleotide, within the following parameters:
- Up to 8 LNA bases
- Deprotected, desalted and purified by PAGE or RP-HPLC
- Available in lengths of 7 to 40 mers
- Delivered dried in individual, opaque tubes
- Shipped in 7 working days of receiving your order, pending successful QC validation*
|
Guaranteed yields of LNA dual-labeled fluorogenic probes and LNA molecular beacons
|
|
Guaranteed yield (OD)
|
Approximate yield (nmols**)
|
Estimated assays (reactions***)
|
|
1
|
5
|
500
|
|
3
|
15
|
1,500
|
|
5
|
25
|
2,500
|
|
10
|
50
|
10,000
|
** Estimate 1 OD = 5 nmols = 30 µg, for a 20 mer oligo *** Estimated number of assays based on a 50 µl reaction volume. Please enquire for alternative quantities.
|
Labels for LNA dual-labeled fluorogenic probes and LNA molecular beacons
|
|
5'-end reporter
|
3'-end quencher
|
|
6-FAM™, HEX™ or TET™
|
TAMRA™, DABCYL, BHQ™-1 or BHQ-2
|
Please inquire about optional dye labels.
Figure 1: LNA dual-labeled fluorogenic probes vs. TaqMan® MGB probes in 5' nuclease PCR assays. (Amplification of Staphylococcal enterotoxin gene in a Staphylococcus aureus strain11).
Key
|
|
LNA dual-labeled fluorogenic probe |
|
|
TaqMan MGB probe |
OD = optical density
* Please check with your Sigma Custom Products representative for local delivery times.
Specifications are subject to change.
|
LNA—Notice of limited rights: Sigma, Sigma-Life Science are only licensed to sell Dual Labeled Fluorescent Probes, LightCycler Probes, Scorpion Probes and Molecular Beacons containing LNA (Locked Nucleic Acids) for use in Real-Time PCR.
|