Sigma-Aldrich
Life Science > Custom Oligos > Custom DNA Probes > Product Lines > LNA Probes > LNA LightCycler® Probes
Dual-Labeled DNA Probes
LNA LightCycler® Probes
For your convenience, LNA LightCycler probes are available at a fixed price per oligonucleotide, within the following parameters:
  • Up to 6 LNA bases
  • Deprotected, desalted and purified by RP-HPLC
  • Available in lengths of 7 to 40 mers
  • Delivered dried in individual, opaque tubes
  • Shipped in 6 working days of receiving your order, pending successful QC validation*
Guaranteed yields of LNA LightCycler probes
Guaranteed yield (OD)
Approximate yield (nmols)
Estimated assays (reactions)
0.1
0.3
100
0.25
1
300
1.5
3
1,000
Please inquire for alternative quantities.
Labels and modifications for LNA LightCycler probes
LNA Probe I
LNA Probe 2
3'-end donor fluorophore
3'-end
5'-end acceptor fluorophore
Labels and modifications
LightCycler Fluorescein
Phosphate
LightCycler Red 640
Note: Fluorescent signaling occurs during the annealing step of Q.PCR.
Please inquire for optional dye labels.
Figure 1: Melting curve profiles for Human Cystic Fibrosis SNP G542X demonstrate the superior discrimination power of LNA LightCycler probes over DNA LightCycler probes.
Figure 1a: A set of DNA LightCycler probes (A-25; D-22)
PP_LNAFP_Figure_1a
Key
I Wild-type template
II Heterozygous template
III Mutant-type template
IV Control without template
Figure 1b: A set of LNA LightCycler probes (A-16; D-16)
PP_LNAFP_Figure_1b
Key
I Wild-type template
II Heterozygous template
III Mutant-type template
Note that DNA LightCycler probes do not pick up on the presence of the homozygous mutant, whereas the LNA LightCycler probes are able to identify the presence of all three template types, including the homozygous mutant.
* Please check with your Sigma-Proligo representative for local delivery times.
Specifications are subject to change.