Sigma-Aldrich
Life Science > Custom Oligos > Custom DNA Probes > Product Lines > LNA Probes > Spectral Properties
Dual-Labeled DNA Probes
Spectral Properties
The table below details the standard dyes and quenchers available for the LNA fluorescent probes, together with their excitation and emission data. Please enquire for any other labels.
Quenchers
Maximum absorbance (nm)
Maximum emission
(nm)
BHQ-1 Dark Quencher
535
None
BHQ-2 Dark Quencher
579
None
DABCYL
453
None
TAMRA
550
576
Quenchers
Maximum absorbance (nm)
Maximum emission
(nm)
6-FAM
495
517
HEX
537
553
LightCycler Fluorescein
495
520
LightCycler Red 640
625
640
TET
521
538