Life Science > Custom Oligos > Custom DNA Probes > Product Lines > Molecular Beacons
Custom DNA Probes
Molecular Beacons
Molecular Beacons are structured probes that are highly sensitive, sequence-specific and used for sequence detection in real-time qPCR and in vitro studies.
Choose Molecular Beacons for:
  • End-point genotyping
  • In vitro quantification or detection studies
  • Multiplexing
  • SNP detection
  • Allelic discrimination
  • Pathogen detection
Benefits of Using Molecular Beacons include:
  • Increased specificity
  • Probe preserved during the reaction
Add LNA to Your Probe for:
  • Increased thermal stability and hybridization specificity
  • Greater accuracy in gene quantitation and allelic discrimination
  • Easier and more sensitive probe designs for problematic target sequences
back to top
How Molecular Beacons Work
A Molecular Beacon is a single-stranded bi-labeled fluorescent probe held in a hairpin-loop conformation (around 20 to 25 nt) by complementary stem sequences (around 4 to 6 nt) at both ends of the probe. The 5' and 3' ends of the probe contain a reporter dye and a quencher dye, respectively. The loop is a single-stranded DNA sequence complementary to the target sequence. The close proximity of the reporter and quencher dyes causes the quenching of the natural fluorescence emission of the reporter dye. The structure and mechanism of a Molecular Beacon is shown below.
How Molecular Beacons Work Image
Molecular Beacons hybridize to their specific target sequence causing the hairpin-loop structure to open and separate the 5' end reporter dye from the 3' end quencher dye. As the quencher dye is no longer in proximity to the reporter dye, fluorescence emission takes place. The measured fluorescence signal is directly proportional to the amount of target DNA.
Product Features include:
  • Available in lengths of 15–40 bases
  • Simple pricing – HPLC purification and base charges are included
  • Stringent quality control including electrospray mass spectroscopy
  • Shipped within 5–6 business days for 6-FAM™, HEX™, or TET™ labeled
  • Shipped in an amber tube to protect dye integrity
  • Custom formats available (normalization, special plates, etc.)
Sigma's probes are provided in a format to simplify your experimental planning.
back to top
Guaranteed Yields
Guaranteed
OD Yield
Approx. No.
of nmoles
Approx. No.
of µg
Approx. No.
of Reactions*
1 3 32 600
3 9 96 1,800
5 15 160 3,000
10 30 320 6,000
*Estimate is based on 3 nmoles or 32 µg for 1 OD and 200 nM in 25 µl reaction (5.0 pmol/reaction). Estimate is based on an average sequence length of 30 bases.
The most common fluorophore and quencher combinations are listed below:
Dye Max.
EX (nm)
Max.
EM (nm)
Compatible
Quencher
6-FAM™ 494 515 BHQ-1, DABCYL
Fluorescein 495 520 BHQ-1, DABCYL
JOE™ 520 548 BHQ-1, DABCYL
TET™ 521 536 BHQ-1, DABCYL
HEX™ 535 555 BHQ-1, DABCYL
Cy3® 550 570 BHQ-2, DABCYL
ROX™ 573 602 BHQ-2, DABCYL
Texas Red® 583 603 BHQ-2, DABCYL
Cy5® 651 674 BHQ-3, DABCYL
Cy5.5® 675 694 BHQ-3, DABCYL
Sigma® is a licensed supplier of a variety of dyes and quenchers and continually adds to its portfolio of new chemistries. For assistance in the design of your probe and/or assays, submit your request at sigma.com/designmyprobe.
back to top
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Custom Oligos DNA