Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Epigenetics > Imprint® ChIP Kit  > Imprint® ChIP Validated Antibodies
Epigenetics
Imprint® ChIP Validated Antibodies
Chromatin structure plays a fundamental role in regulating processes that take place on the DNA, such as transcription, replication, and recombination, all of which occur on nucleosomal templates. Therefore, mapping the position of histones selectively modified and histone variants or non-histone components of chromatin in specific DNA sequences, can provide valuable insights into how these proteins (and their modifications) function in a chromatin context.
Chromatin immunoprecipitation (ChIP) is a powerful tool for identifying proteins, including histone proteins and non-histone proteins, associated with specific regions of the genome by using specific antibodies that recognize a specific protein or a specific modification of a protein. The method is based on the principle that formaldehyde reacts with primary amines located on amino acids and the bases on DNA or RNA molecules, forming a covalent crosslink between the specific protein to the DNA on which they are situated.
Once the DNA binding proteins are crosslinked, the cells are lysed and the crude cell extracts are sonicated to shear the DNA to a smaller size. The protein:DNA complex is immunoprecipitated using an antibody against the protein of interest. The DNA protein cross-links are reversed by heating and the proteins removed by treatment with proteinase K. The DNA portion of the complex is then purified and identified by PCR using specific primers to the suspected binding region.
Successful chromatin immunoprecipitation requires an effective, specific, and high quality antibody. Not all antibodies work in ChIP as the antigen epitope may be blocked or altered once cross-linked. Sigma’s Imprint® brand of antibodies are validated in ChIP application and lot tested each time for successful chromatin immunoprecipitation experiment.
Product No. Description
SAB4800001 Imprint Monclonal Anti-5-methylcytosine (33D3) antibody produced in mouse
SAB4800002 Imprint Monclonal Anti-5-hydroxymethylcytosine antibody produced in rat
SAB4800003 Imprint Anti-H3K4me3, (N-terminal) antibody produced in rabbit
SAB4800004 Imprint Anti-H3K9me3 antibody produced in rabbit
SAB4800005 Imprint Anti-H3K9me2 antibody produced in rabbit
SAB4800006 Imprint Anti-H3K36me3 antibody produced in rabbit
SAB4800007 Imprint Anti-H3K9/14ac antibody produced in rabbit
SAB4800008 Imprint Anti-H3K4me2, (N-terminal) antibody produced in rabbit
SAB4800009 Imprint Anti-H3Pan antibody produced in rabbit
SAB4800010 Imprint Anti-H3K4me1, (N-terminal) antibody produced in rabbit
SAB4800011 Imprint Monclonal Anti-TBP, (N-terminal) antibody produced in mouse
SAB4800012 Imprint Anti-MeCP2 antibody produced in rabbit
SAB4800013 Imprint Anti-TIP5 antibody produced in rabbit
SAB4800014 Imprint Anti-HDAC1, (C-terminal) antibody produced in rabbit
SAB4800015 Imprint Anti-H3K27me3 antibody produced in rabbit
SAB4800016 Imprint Anti-H3K9me1 antibody produced in rabbit
SAB4800017 Imprint Anti-H4K20me3 antibody produced in rabbit
SAB4800018 Imprint Monclonal Anti-H3K9me3 antibody produced in mouse
SAB4800019 Imprint Monclonal Anti-HDAC1, (C-terminal) antibody produced in mouse
SAB4800020 Imprint Monclonal Anti-H4K20me1 antibody produced in mouse
SAB4800021 Imprint Anti-H3K36me1 antibody produced in rabbit
SAB4800022 Imprint Anti-H3K4ac, (C-terminal) antibody produced in rabbit
SAB4800023 Imprint Anti-H3S10p antibody produced in rabbit
SAB4800024 Imprint Anti-H4Pan antibody produced in rabbit
SAB4800025 Imprint Anti-H3K27me3 antibody produced in rabbit
SAB4800026 Imprint Anti-H3K4me3, (C-terminal) antibody produced in rabbit
SAB4800027 Imprint Anti-H3K9me3 antibody produced in rabbit
SAB4800028 Imprint Anti-H3K36me3 antibody produced in rabbit
SAB4800029 Imprint Anti-H3K79me2 antibody produced in rabbit
SAB4800030 Imprint Monclonal Anti-H3K9me3 antibody produced in rabbit
SAB4800031 Imprint Anti-H3K9me1 antibody produced in rabbit
SAB4800032 Imprint Monclonal Anti-Ty1 antibody produced in mouse
SAB4800033 Imprint Anti-H3K27me3S28p antibody produced in rabbit
SAB4800034 Imprint Anti-MBD1 antibody produced in rabbit
SAB4800035 Imprint Anti-H4K8ac antibody produced in rabbit
SAB4800036 Imprint Anti-H3S10p antibody produced in rabbit
SAB4800037 Imprint Anti-H3K36me2 antibody produced in rabbit
SAB4800038 Imprint Anti-H3R17me2 antibody produced in rabbit
SAB4800039 Imprint Monclonal Anti-H4K20me1 antibody produced in mouse
SAB4800040 Imprint Monclonal Anti-ERa antibody produced in mouse
SAB4800041 Imprint Monclonal Anti-GR antibody produced in mouse
SAB4800042 Imprint Anti-H2Bpan antibody produced in rabbit
SAB4800043 Imprint Anti-H3K79me1 antibody produced in rabbit
SAB4800044 Imprint Anti-H3K9acS10p antibody produced in rabbit
SAB4800045 Imprint Anti-H3K9me3S10p antibody produced in rabbit
SAB4800046 Imprint Anti-H4K8ac antibody produced in rabbit
SAB4800047 Imprint Anti-RARa antibody produced in rabbit
SAB4800048 Imprint Monclonal Anti-Ago antibody produced in mouse
SAB4800049 Imprint Monclonal Anti-5-hydroxymethylcytosine antibody produced in mouse
SAB4800051 Imprint Anti-5-hydroxymethylcytosine antibody produced in rabbit
Products to Support the Epigenetics Workflow using this Kit
DNA Extraction and Amplification
Product No. Description
G1N10 GenElute™ Mammalian Genomic DNA Miniprep Kit
NA2010 GenElute Blood Genomic DNA Kit
WGA1 GenomePlex® Whole Genome Amplification (WGA) Kit
WGA2 GenomePlex Complete Whole Genome Amplification (WGA) Kit
WGA3 GenomePlex WGA Reamplification Kit
WGA4 GenomePlex Single Cell Whole Genome Amplification Kit
WGA5 GenomePlex Tissue Whole Genome Amplification Kit
Chromatin Immunoprecipitation Kits
For highly expressed proteins and histone modifications
Product No. Description
CHP1-24RXN
CHP1-96RXN
Imprint Chromatin Immunoprecipitation Kit
For low abundance proteins, transcription factors and ChIP sequencing
Product No. Description
CHP2-24RXN
CHP2NC-48RXN
Imprint Ultra Chromatin Immunoprecipitation Kit
Imprint® Ultra Chromatin Immunoprecipitation Kit without controls
CHROP-15RXN Imprint Ultra Chromatin Optimization Kit