Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Epigenetics > DNA Purification
Epigenetics
DNA Purification
Sigma provides a variety of kits that may be used to isolate DNA to begin your Epigenetic workflow. Our GenElute™ product line provides a simple and convenient way to purify high-quality Genomic DNA from mammalian cells and tissues, blood, and plants. Each kit utilizes a spin column format to rapidly extract Genomic DNA free of contaminants. The purified product is ideal for use in DNA Methylation analysis.
GenElute Mammalian
Genomic DNA Miniprep Kit
  GenElute Blood
Genomic DNA Kit
DNA yields of 25-30 µg can be isolated from up to 2x106 cells or 25mg of tissue in less than 40 minutes. All reagents, columns and tubes provided.
G1N10      10 Purifications
G1N70      70 Purifications
G1N350    350 Purifications
For additional information visit the GenElute Mammalian DNA Miniprep Kit page
  DNA may be isolated from fresh or aged blood in less than 40 minutes. All contaminants are removed by provided prewash solution, an Rnase solution is provided as well.
NA2000    10 Purifications
NA2010    70 Purifications
NA2020    350 Purifications
For additional information visit the GenElute Blood Genomic DNA Kit page
GenElute Plant
Genomic DNA Miniprep Kit
Up to 100 mg fresh or 20 mg of freeze-dried plant tissue can result in DNA yields of 20 µg per prep. Individual preps take no longer than 40 minutes and do not require Rnase A digestion.
G2N10      10 Purifications
G2N70      70 Purifications
G2N350    350 Purifications
For additional information visit the GenElute Plant Genomic DNA Kit page
Epigenetics Overview