Epigenetics
Sperm Nuclei Prep
cshlp-small Sigma® has prepared product lists for commonly used protocols from Cold Spring Harbor Protocols.
View the entire protocol on BioSupplyNet.com.
View the entire protocol,"Generation of Transgenic Xenopus laevis" at BioSupplynet.com
Shoko Ishibashi1, Kristin L. Kroll2, and Enrique Amaya1,3
1 The Healing Foundation Centre, Faculty of Life Sciences, University of Manchester, Manchester M13 9PT, United Kingdom
2 Department of Molecular Biology and Pharmacology, Washington University School of Medicine, St. Louis, MO 63110, USA
3 Corresponding author (enrique.amaya@manchester.ac.uk)
Generation of Transgenic Xenopus laevis: I. High-Speed Preparation of Egg Extracts. Please cite as CSH Protocols; 2007; doi:10.1101/pdb.prot4838.
ABSTRACT
Manipulating genes specifically during later stages of amphibian embryonic development requires fine control over the time and place of expression. These protocols describe an efficient nuclear-transplantation-based method of transgenesis developed for Xenopus laevis. The approach enables stable expression of cloned gene products in Xenopus embryos. Because the transgene integrates into the genome prior to fertilization, the resulting embryos are not chimeric, eliminating the need to breed to the next generation to obtain nonmosaic transgenic animals. The procedure is based on restriction-enzyme-mediated integration (REMI) and can be divided into three parts: (I) high-speed preparation of egg extracts, (II) sperm nuclei preparation, and (III) nuclear transplantation. This protocol describes the method for the high-speed preparation of egg extracts. Briefly, a crude, cytostatic factor (CSF)-arrested egg extract (i.e., cytoplasm arrested in meiotic metaphase) is prepared. These extracts are driven into the interphase stage of the cell cycle by addition of calcium, and high-speed centrifugation is performed to obtain a purer cytoplasmic fraction. This fraction promotes swelling of sperm nuclei, but does not promote DNA replication. By adding the egg extract to the reaction, the sperm chromatin partially decondenses, facilitating integration of plasmid DNA into the genome.
Related Information:
For details on basic Xenopus procedures used in this protocol, please see Handling Xenopus laevis Adults, Inducing Ovulation in Xenopus laevis, Xenopus laevis Egg Collection, and Dejellying Xenopus laevis Embryos.
Products Available for this Protocol
Protocol Material Description Product #  Product Name Add to Cart
10% bovine serum albumin A7906 Albumin from bovine serum, pH ~7 (1% in 0.15 M NaCl), ≥98% (agarose gel electrophoresis), lyophilized powder
Hoechst No. 33342 B2261 bisBenzimide H 33342 trihydrochloride, ≥98% (HPLC and TLC)
NaOH 655104 Sodium hydroxide, reagent grade, 97%, powder
Human chorionic gonadotropin (HCG) C0434 Chorionic gonadotropin human, from human pregnancy urine, lyophilized powder
L-α-lysolecithin, egg yolk L4129 L-α-Lysophosphatidylcholine from egg yolk, ~99%, Type I, powder
CaCl2 C2661 Calcium chloride, plant cell culture tested, anhydrous
KCl P9541 Potassium chloride, for molecular biology, ≥99%
MgCl2 M4880 Magnesium chloride, insect cell culture tested
NaCl S3014 Sodium chloride, for molecular biology, ≥98% (titration)
Dithiothreitol D5545 DL-Dithiothreitol, SigmaUltra, >99% (titration)
EDTA (pH 8.0) E7889 Ethylenediaminetetraacetic acid disodium salt solution, for molecular biology, 0.5 M
HEPES H4034 HEPES, Biotechnology Performance Certified, ≥99.5% (titration), cell culture tested
Spermidine trihydrochloride S2501 Spermidine trihydrochloride, ≥98% (TLC)
Spermine tetrahydrochloride S1141 Spermine tetrahydrochloride, for molecular biology
Sucrose S7903 Sucrose, SigmaUltra, pH 5.5–7.5 (20 °C, 1 M in H2O), >99.5% (GC)
Pregnant mare serum gonadotropin (PMSG) G4877 Gonadotropin from pregnant mare serum, ≥1,000 IU/mg
Glycerol G5516 Glycerol, for molecular biology, ≥99%
Tricaine methanesulfonate E10521 Ethyl 3-aminobenzoate methanesulfonate, 98%
Sodium bicarbonate S7277 Sodium bicarbonate, for molecular biology, 99.7–100.3%
Chymostatin C7268 Chymostatin, microbial
DMSO D2438 Dimethyl sulfoxide, Biotechnology Performance Certified, sterile-filtered, hybridoma tested, meets EP, USP testing specifications
Leupeptin L9783 Leupeptin hydrochloride, microbial, ≥90% (HPLC)
Pepstatin P5318 Pepstatin A, microbial, ≥90% (HPLC)
L-cysteine hydrochloride
1-hydrate
C121800 (R)-(+)-Cysteine hydrochloride hydrate
EGTA E3889 Ethylene glycol-bis(2-aminoethylether)-N,N,N',N'-tetraacetic acid, for molecular biology, ≥97%
ATP A7699 Adenosine 5'-triphosphate disodium salt, SigmaUltra, ≥99%
Creatine phosphate 27920 Sodium creatine phosphate dibasic tetrahydrate, BioChemika, ≥98.0% (NT)
Back to Top
View all available product lists for Cold Spring Harbor Laboratory Protocols
Product Association Disclaimer: The Sigma-Aldrich products listed for this specific protocol were selected either to match or to supplement the products listed within the actual protocol. The products/reagents from Sigma-Aldrich have been qualified for usage, but may not have been validated for this specific application. Please refer to the detailed product description on the usage of specific products of interest.