Return to Web Version Sigma-Aldrich
Lenti GoClone
Complete Reporter System for Human miRNA Target Validation
MISSION® 3′UTR Lenti GoClone powered by SwitchGear Genomics™
Non-Coding RNA
SwitchGear Genomics Logo
An Ideal Functional Tool for Human miRNA Target Validation
Validate miRNA targets and study post-transcriptional gene regulation with lentiviral 3′UTR reporters.
Sigma® Life Science has partnered with SwitchGear Genomics to offer the MISSION 3’UTR Lenti GoClone system. The new system utilizes viral vectors to deliver human 3′UTRs fused to SwitchGear’s novel RenSP reporter gene. LightSwitch Luciferase Assay Reagents have been designed specifically for use with RenSP and deliver maximum sensitivity, dynamic range, and convenience.
Find Your Lenti GoClone
Benefits of using the MISSION 3′UTR Lenti GoClone Products
Workflow
MISSION® 3'UTR Lenti GoClone
  1. Transduce a MISSION 3'UTR Lenti GoClone into your cell line of choice. 
  2. A constitutive promoter drives production of a hybrid luciferase (RenSp)-human 3’UTR transcript. The RenSP luciferase produces luminescence when subjected to luciferase substrate
  3. Transfect miRNA mimic.
  4. If the inserted miRNA recognizes and binds to the specific 3’UTR, the miRNA will affect the transcript stability and/or translational efficiency and there will be a reduction in luminescence.
  5. Add luciferase substrate and measure luminescence by use of luminometer.