Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Functional Genomics & RNAi
MicroRNA
mirPremier™ microRNA Isolation Kit
More rapid, more pure, less hazard, less hands on.
The mirPremier microRNA Isolation Kit complements the already robust MISSION® RNAi product portfolio which includes a broad choice of MISSION siRNA and shRNA products and services such as libraries, mRNA detection reagents, antibodies and AQUA peptides for protein level detection as well as an entire workflow of supporting reagents (cell culture, cell biology compounds and assays, transfection reagents, and much more). For more information, visit the Functional Genomics & RNAi homepage.
MicroRNA (miRNA) are a class of small RNA molecules, about 21 nucleotides in length that regulate gene expression in a variety of ways, including translational repression, mRNA cleavage and deadenylation. Developed by Sigma, the mirPremier microRNA Isolation Kit is a new member of the MISSION® RNAi product family that provides a rapid and efficient method for purifying and enriching miRNA along with other small RNAs, allowing researchers to obtain high-quality miRNA directly from cells or tissues as rapidly and simply as a total RNA prep.
  • Optimized for purification of microRNA and other small RNA directly from diverse biological sources
  • Rapidly purifying and enriching miRNA in 30 minutes for downstream applications
  • High-purity miRNA with no detectable large RNA
  • No hazardous organic extractions
Purification of miRNA from diverse biological sources
Using mirPremier microRNA Isolation Kit, researchers can purify miRNA/small RNA from diverse biological sources, including mammalian cell cultures, animal tissues, microbial cultures and plant tissues (Figure 1). mirPremier microRNA Isolation Kit also allows researchers to purify miRNA/small RNA with high yield from "difficult-to-extract" tissues, such as grape leaf (Figure 2). In addition, the kit can be used to isolate total RNA if messenger RNA or other large RNAs are of interest.
Figure 1
Figure 1
miRNA samples purified with mirPremier microRNA Isolation Kit from diverse biological materials. Each sample was analyzed on an Agilent Bioanalyzer using an RNA Nano Kit.
Figure 2
Figure 2
Isolation of small RNA from a difficult-to-extract plant tissue — grape leaf — using different microRNA isolation kits. Grape leaf tissues were first ground into a fine powder in liquid nitrogen and then 100 mg was extracted with different microRNA isolation kits. Each purified sample was analyzed with 5% of the total recovered volume on 4% agarose gel containing ethidium bromide. The DNA marker is an MspI digest of puc18 (mw- 67, 89, 110, 147, 190, 242, 353, 404, 49, and 501 bases). Yeast tRNA was loaded with 250 ng per lane.
Rapid purification and enrichment of miRNA within 30 minutes
Timely extraction of miRNA from cells or tissues is critical for downstream applications. mirPremier™ microRNA Isolation Kit provides a robust protocol that enables scientists to purify miRNA within 30 minutes, making it an ideal solution for rapid preparation of miRNA (Figures 3 and 4).
Figure 3
Figure 3
Comparison of times for miRNA purification.
Figure 4
Figure 4
Comparison of small RNA purification workflow.
Superior quality of miRNA/small RNA without any detectable large RNA
Unlike other miRNA purification methods, mirPremier microRNA Isolation Kit yields high-purity miRNA/small RNA, containing no detactable large RNA (Figure 5).
Figure 5
Figure 5
miRNA quality comparison between Sigma mirPremier microRNA Isolation Kit (A) and competitor A miRNA purification kit (B). Two peaks on the right of Fig. 5B are contaminating rRNA. Each sample was purified from 40 mg mouse liver tissues and analyzed with 1% of the total recovery by Agilent Bioanalyzer using an RNA Nano kit.
Contains no hazardous organic extractions
The mirPremier microRNA Isolation Kit employs a novel purification chemistry to isolate miRNA and other small RNAs in a simplified and streamlined manner, without using hazardous organic extractions such as phenol and chloroform.
Quality and Service with each and every lot
Like other Sigma life science products, the mirPremier microRNA Isolation Kit is backed by the quality production standards, customer service, technical support, and other extensive offerings that make Sigma the most trusted name in life science technology.
Ordering Information
Product No. Description Add to Cart
SNC10 mirPremier microRNA Isolation Kit (10 preparations)
SNC50 mirPremier microRNA Isolation Kit (50 preparations)
References
  1. Ausubel, F. M. et al. Current Protocols in Molecular Biology; John Wiley & Sons: New York, 1995; Sections 4.1–4.10.
  2. Farrell, Robert E., Jr. RNA Methodologies, 2nd Edition; Academic Press: New York, 1998; pp 37–53.
  3. Sambrook, J. et al. Molecular Cloning: A Laboratory Manual, 2nd edition; Cold Spring Harbor Laboratory Press: Plainview, New York, 1989; pp 7.3–7.5.
Back to Top