Life Science > Functional Genomics & RNAi > MISSION® esiRNA for RNAi Screening in Mammalian Cells
Life Science
MISSION® esiRNA for RNAi Screening in Mammalian Cells
Endoribonuclease-prepared siRNAs (esiRNAs) are pools of siRNAs resulting from cleavage of long double-stranded RNA (dsRNA) with Escherichia coli RNase III.
Our MISSION esiRNA products are ideal for simple, cost-effective RNAi screening. MISSION esiRNA are available for individual mouse and human genes, custom plated and genome scale libraries.
  • Lower off-target effects than single or pooled siRNAs
  • Guaranteed ≥70% knockdown at the mRNA level
  • Lowest cost genome scale RNAi screening tool
  • Provides quick primary screens
  • >16,000 human and >14,000 mouse targets available
Browse a complete list of esiRNA
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page