Return to Web Version Sigma-Aldrich
Life Science
MISSION® esiRNA for RNAi Screening in Mammalian Cells
Endoribonuclease-prepared siRNAs (esiRNAs) are pools of siRNAs resulting from cleavage of long double-stranded RNA (dsRNA) with Escherichia coli RNase III.
Our MISSION esiRNA products are ideal for simple, cost-effective RNAi screening. MISSION esiRNA are available for individual mouse and human genes, custom plated and genome scale libraries.
Search now for your esiRNA of interest
Enter your gene of interest into the box below.
Or
Browse a complete list of esiRNA
For Library Inquiries:
In the United States, please contact your local Sigma Representative or email us at
acctreps-hou@sial.com.
For all other countries, please contact your local Sigma Representative or email us at MissionRNAi@sial.com.