Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
siRNA
esiRNA/esiFLEX/Non-coding
Save 15% with Promo Code SGF
Global Customers
Expires June 30, 2015
MISSION esiRNA
Eupheria Biotech
On this page:
MISSION esiRNA
Benefits
esiRNA Targeting Long Non-coding RNA
esiRNA Screening Video Tutorial
esiRNA FAQ (Frequently Asked Questions)
MISSION esiRNA Offering
Ordering Information
esiRNA Controls
References
MISSION® esiRNA
MISSION esiRNA provides RNAi researchers with a proven, cost-effective, and simple way to perform RNAi screens. MISSION esiRNAs are endoribonuclease-prepared siRNA pools comprised of a heterogeneous mixture of siRNAs that all target the same mRNA sequence. These multiple silencing triggers lead to highly specific and effective gene silencing with lower off-target effects than single or pooled siRNAs.
Product Offerings
MISSION esiRNA and MISSION esiFLEX are available individually, as custom arrayed panels, or genome scale libraries that target both human and mouse genes.
Product # esiRNAs Coding Genes # esiRNAs Long Non- coding Scale (µg) Format Features and Ordering Information
Individual esiRNAs 16,744 (human)
14,068 (mouse)
1,761 (human)
643 (mouse)
20 / 50 Single Tubes
96/384-well plates
Easy Online Ordering
Search now
esiFLEX 16,744 (human)
14,068 (mouse)
1,761 (human)
643 (mouse)
1 / 2.5 / 5 96 / 384-well plates Custom-array. Cherry-pick the library.
Contact us
Libraries 16,744 (human)
14,068 (mouse)
1,761 (human)
643 (mouse)
1 / 2.5 / 5 384-well plates Pre-arrayed library.
Contact us
esiOPEN Custom
All species
Custom 20 / 50 Single tubes Custom sequence (DEQOR optimization).
Contact us
esiSEC 16,744 (human)
14,068 (mouse)
1,761 (human)
643 (mouse)
20 / 50 Single tubes Secondary independent esiRNA for validation of primary esiRNA.
Contact us
Learn more about our coding esiRNAs.
Learn more about our long non-coding esiRNAs.
Spend your time on discovery!
Features and Benefits
  • Lower off target effects than single or pooled siRNAs
  • Guaranteed ≥70% knockdown at the mRNA level
  • Lowest cost genome scale RNAi screening tool
  • >16,000 human and >14,000 mouse targets available
  • Provides quick primary screens
  • High on-target specificity
How are MISSION esiRNAs prepared?
esiRNA or Endoribonuclease-prepared siRNAs are pools of siRNAs resulting from cleavage of long double-stranded RNA (dsRNA) with Escherichia coli RNase III. Please see Figure 1.
Figure 1
: Overview of MISSION esiRNA Production.
Note: This is for the production of an individual esiRNA.
Generation of PCR products
The template for in vitro transcription is generated by PCR amplification of the cDNA from the clone using primers specific for the vector backbone or target-specific primers appended with RNA-Polymerase promoter sequences. The PCR product is sequence verified. The amplified region is selected based on the highest possible number of highly effective siRNA based on the DEQOR siRNA design program. MISSION esiRNA are designed to cover all known transcript variants of the target gene.
In vitro transcription and the production of long dsRNA
The PCR product generated from the cDNA is used for in vitro transcription reactions. The dsRNA is produced utilizing RNA Polymerases and the two single RNA strands are subsequently annealed before digestion.
Enzymatic digestion
The long dsRNA is enzymatically digested to short dsRNAs. This digestion produces complex pools of siRNA-like molecules.
Purification of digested dsRNA
The digestion is then purified to remove any remaining DNA template, unincorporated nucleotides, and dsRNAs longer than approximately 40 bp. This purification step results in MISSION esiRNAs with an average length of 21 bp.
MISSION esiRNAs
Since the starting material is cDNA, each MISSION esiRNA is guaranteed to target a real gene, no more wasted time on assays for "predicted" genes that do not actually exist.
MISSION esiRNA are a heterogeneous mixture of siRNAs that all target the same mRNA sequence. These multiple silencing triggers lead to highly specific and effective gene silencing.
esiRNA video tutorial for RNAi Screening in Mammalian Cells
esiRNA Video
esiRNA FAQ (Frequently Asked Questions)
MISSION esiRNA Offering
MISSION esiRNA and MISSION esiFLEX are available individually, as custom arrayed panels, or genome scale libraries that target both human and mouse genes.
Click here to view additional esiRNA product information.
Browse a complete list of esiRNA.
Individual esiRNA
Our individual esiRNAs are ideal for the biological characterization of single genes. Currently, we offer esiRNAs targeting 16,744 human and 14,068 mouse transcripts at quantities of either 20 µg or 50 µg normalized to 200 ng/µl; larger scales are available upon inquiry. All esiRNAs are designed based on sequence annotations from ENSEMBL database and packaged in single tubes.
Plated esiRNA
esiRNAs provided in 96-well customized plates at quantities of either 20 µg or 50 µg normalized to 200 ng/µl. Currently, we offer esiRNAs targeting 16,744 human and 14,068 mouse transcripts. Plate orders must have ≥10 targets.
esiRNA Libraries
MISSION esiRNA libraries are available for whole genome and long non-coding for human and mouse targets. Our genome scale esiRNA libraries are state-of-the-art RNAi reagents especially suitable for large-scale loss-of-function screening.
Currently, we offer a human library composed of 16,744 esiRNAs and a mouse library with 14,068 esiRNAs. All esiRNAs are designed based on sequence annotations from the ENSEMBL database. The long non-coding RNA libraries are composed of 1,761 (human) and 643 (mouse) esiRNAs. The libraries are delivered in 96-well or 384-well plates with all positions occupied at 1 µg, 2.5 µg, or 5.0 µg quantities normalized to 50 ng/µl. Other quantities, custom arrays or other plate formats are available upon request.
esiFLEX esiRNA
Our esiFLEX sub-genomic collections of esiRNAs provide a convenient small scale option with plate position customization. We provide esiFLEX for human or mouse species and offer 16,744 human and 14,068 mouse esiRNAs in our portfolio. The esiFLEX libraries are provided in 96-well plates at 1 µg, 2.5 µg, or 5 µg quantities normalized to 50 ng/µl. The position of each esiRNA on the plates can freely be chosen. Other quantities, concentrations or plate formats are available upon inquiry. All esiRNAs are designed based on sequence annotations from the ENSEMBL database. Minimum order of 24 targets required.
esiOPEN esiRNA
esiOPEN provides access to fully flexible custom-targeting esiRNAs. Provide any transcript-sequence (minimum 500 bp) regardless of the species and an esiRNA will be synthesized against the best target-region (determined by DEQOR) that lies within the provided sequence to ensure maximum efficiency and specificity. esiOPEN esiRNAs are offered at either 20 µg or 50 µg scales normalized to 200 ng/µl and packaged in single tubes. Larger scales are available upon inquiry.
esiSEC esiRNA
esiSEC provides secondary independent silencing triggers for any primary esiRNA in our portfolio. esiSEC are used for the validation of RNAi-phenotypes induced by our primary esiRNAs. esiSEC esiRNAs are offered at either 20 µg or 50 µg scales normalized to 200 ng/µl and packaged in single tubes. Larger scales are available upon inquiry. All esiRNAs are designed based on sequence annotations from the ENSEMBL database.
esiRNA Ordering Information
Individual esiRNAs
Easily find your MISSION esiRNA target by inserting your gene of interest into the search box above and add selected esiRNA to your cart.
For all other esiRNA product offerings:
Controls
  • Negative controls: RLUC, FLUC, and eGFP MISSION esiRNAs
  • Positive control: eg5 (KIF11) MISSION esiRNA (strong mitotic arrest/viability phenotype)
  • MISSION esiRNA targeting eGFP can also be used as a positive control for knockdown in cells expressing eGFP
  • Validated human MISSION esiRNA—Many common gene targets, including LAMIN A, PLK4, and AURKB, have been validated for >70% mRNA knockdown. Validated MISSION esiRNA are suitable for transfection optimization and as positive controls
  • qPCR validation data
  • Western Blot validation data
Validated Human MISSION esiRNA
Gene Name esiRNA Product No. Gene Name esiRNA Product No. Gene Name esiRNA Product No.
ANAPC1 EHU029991 DCTN2 EHU086581 PLK4 EHU001691
ANAPC10 EHU108801 ETN3 EHU040581 RAD21 EHU108911
ANAPC5 EHU069691 H3F3A EHU127091 RCD-8 EHU094431
AURKB EHU001471 INCENP EHU001891 SEC13L1 EHU152131
c14orf10 EHU143981 KIF11 EHU019931 SEH1L EHU049161
CENPA EHU098081 KIF23 EHU066121 SUV39H2 EHU093651
CEP192 EHU026711 LMNA EHU063791 TACC3 EHU063201
CETN2 EHU137031 MAD2L1 EHU074611 TUBG1 EHU056411
ch-TOG EHU078221 NEK2 EHU109951 TUBGCP3 EHU073291
CLASP1 EHU028941 NUMA EHU059141 TPX2 EHU093011
CP110 EHU007261 NUP37 EHU048011    
References
MISSION esiRNA — Proven RNAi Screening Tool
Roguev, A. et al. Quantitative genetic-interaction mapping in mammalian cells. Nature Methods 10, 432–437 (2013).
Abbasi, M., Lavasanifar, A. & Uludag, H. Recent attempts at RNAi-mediated P-glycoprotein downregulation for reversal of multidrug resistance in cancer. Medicinal research reviews 33, 33-53 (2013).
Alvarez-Calderon, F., Gregory, MA. & DeGregori, J. Using functional genomics to overcome therapeutic resistance in hematological malignancies. Immunologic research 55, 100-15 (2013).
Kim, N. & Song, K. KIFC1 is essential for bipolar spindle formation and genomic stability in the primary human fibroblast IMR-90 cell. Cell structure and function (2013).
Ngondo-Mbongo, RP., Myslinski, E., Aster, JC. & Carbon, P. Modulation of gene expression via overlapping binding sites exerted by ZNF143, Notch1 and THAP11. Nucleic acids research (2013).
Vella, P. et al. Tet Proteins Connect the O-Linked N-acetylglucosamine Transferase Ogt to Chromatin in Embryonic Stem Cells. Molecular cell 49, 645-56 (2013).
Calegari, F. et al. Tissue-specific RNA interference in postimplantation mouse embryos with endoribonuclease-prepared short interfering RNA. PNAS, 99, 14236-14240 (2002).
Lin, Z. et al. (2011) Reduced Level of Ribonucleotide Reductase R2 Subunits Increases Dependence on Homologous Recombination Repair of Cisplatin-Induced DNA Damage. Mol Pharmacol doi:10.1124/mol.111.074708. Abstract
Krastev, D.B. et al. A systematic RNAi synthetic interaction screen reveals a link between p53 and snoRNP assembly. Nat Cell Biol 13, 809-18 (2011). Abstract
Nitzsche, A. et al. RAD21 cooperates with pluripotency transcription factors in the maintenance of embryonic stem cell identity. PLoS One 6, e19470 (2011). Abstract
Leushacke, M. et al. An RNA interference phenotypic screen identifies a role for FGF signals in colon cancer progression. PLoS One (2011), accepted for publication.
Slabicki M., et al. (2010) A Genome-Scale DNA Repair RNAi Screen Identifies SPG48 as a Novel Gene Associated with Hereditary Spastic Paraplegia. PLoS Biol 8(6): e1000408. doi:10.1371/journal.pbio.1000408. Paper (2.22 Mb PDF)
Collinet, et al. Systems survey of endocytosis by multiparametric image analysis. Nature 464, 243-249 (2010). Abstract
Theis, M. et al. Comparative profiling identifies C13orf3 as a component of the Ska complex required for mammalian cell division. EMBO J. 28, 1453-65 (2009). Abstract
Ding, L. et al. A Genome-Scale RNAi Screen for Oct4 Modulators Defines a Role of the Paf1 Complex for Embryonic Stem Cell Identity. Cell Stem Cell. 9, 403-15 (2009). Abstract
Kittler, R. et al. Genome-scale RNAi profiling of cell division in human tissue culture cells. Nat. Cell Biol. 9, 1401-12 (2007). Abstract
Kittler, R. et al. An endoribonuclease-prepared siRNA screen in human cells identifies genes essential for cell division. Nature 432, 1036-40 (2004). Abstract
Additional esiRNA Literature References
Stewart, G. S. et al. The RIDDLE syndrome protein mediates a ubiquitin-dependent signaling cascade at sites of DNA damage. Cell 136, 420-34 (2009). Abstract
Lawo, S. et al. HAUS, the 8-Subunit Human Augmin Complex, Regulates Centrosome and Spindle Integrity. Curr Biol., 19, 816-26 (2009). Abstract
Fazzio, T. G. et al. An RNAi screen of chromatin proteins identifies Tip60-p400 as a regulator of embryonic stem cell identity. Cell 2008 134, 162-74 (2008). Abstract
Konstantinova, I. et al. EphA-Ephrin-A-mediated beta cell communication regulates insulin secretion from pancreatic islets. Cell 129, 359-70 (2007). Abstract
Nikolova, G. et al. The vascular basement membrane: a niche for insulin gene expression and Beta cell proliferation. Dev Cell. 10, 397-405 (2006). Abstract
Kittler, R. et. al. RNA interference rescue by bacterial artificial chromosome transgenesis in mammalian tissue culture cells. Proc. Natl. Acad. Sci. U.S.A. 102, 2396-401 (2005). Abstract
Liu, W. Y. et al. Efficient RNA interference in zebrafish embryos using synthesized with SP6 RNA polymerase. Dev. Growth Differ. 47(5), 323-31 (2005). Abstract
Yang, D. et al. Short RNA duplexes produced by hydrolysis with Escherichia coli RNase III mediate effective RNA interference in mammalian cells. Proc. Natl. Acad. Sci. U.S.A. 99, 9942-7 (2002). Abstract
Contact Us
For technical questions concerning esiRNAs, please email our technical Service department at techserv@sial.com
MISSION is a registered trademark of Sigma-Aldrich Co. LLC Label License.
back to top